Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00820743_g1
          See other RPS26 GE Assays ›
          Gene Symbol:
          Rps26 LOC100360508 LOC100361854
          Gene Name:
          ribosomal protein S26
          ribosomal protein S26-like
          ribosomal protein S26-like
          Gene Aliases:
          -
          -
          -
          Chromosome Location:
          Chr.7: 3052112 - 3053662 on Build Rnor_6.0
          Chromosome Location:
          Chr.13: 45079857 - 45080300 on Build Rnor_6.0
          Chromosome Location:
          Chr.X: 115495660 - 115496062 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          Assay ID Rn00820743_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rps26
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013224.1 NP_037356.1 3 - 4 338 83
          GenBank mRNA BC061561.1 - 3 - 4 342 83
          FQ210888.1 - 3 - 4 338 83
          FQ211456.1 - 3 - 4 337 83
          FQ216976.1 - 3 - 4 338 83
          FQ217039.1 - 3 - 4 338 83
          FQ217630.1 - 3 - 4 338 83
          FQ220803.1 - 3 - 4 409 83
          FQ221742.1 - 3 - 4 338 83
          FQ221917.1 - 3 - 4 339 83
          FQ222007.1 - 3 - 4 338 83
          FQ222170.1 - 3 - 4 338 83
          FQ222423.1 - 3 - 4 339 83
          FQ222638.1 - 3 - 4 338 83
          FQ223096.1 - 3 - 4 337 83
          FQ223202.1 - 3 - 4 338 83
          FQ223253.1 - 3 - 4 334 83
          FQ223633.1 - 3 - 4 338 83
          FQ224810.1 - 3 - 4 338 83
          FQ229027.1 - 3 - 4 338 83
          FQ233862.1 - 3 - 4 338 83
          FQ233965.1 - 2 - 3 338 83
          FQ234974.1 - 3 - 4 339 83
          FQ235169.1 - 3 - 4 339 83
          FQ235253.1 - 3 - 4 338 83
          X02414.1 - 3 - 4 338 83
          Gene Symbol: LOC100360508
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_002728001.3 XP_002728047.2 1 372 83
          Gene Symbol: LOC100361854
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_002730210.4 XP_002730256.1 1 332 83

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rps26
          Click here to find siRNAs for Gene LOC100360508
          Click here to find siRNAs for Gene LOC100361854

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          biological_process
          negative regulation of RNA splicing
          molecular_function
          RNA binding
          mRNA binding
          structural constituent of ribosome

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P049884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660489 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54892 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P049884 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P049884 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P049884 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11759 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700145614 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37675 - -
          Agilent Rat cDNA Microarray (G4105A) 700145614 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10899870 - -
          Affymetrix Rat Gene 1.0 ST Array 10899871 - -
          Affymetrix Rat Gene 1.0 ST Array 10751768 - -
          Affymetrix Rat Gene 1.0 ST Array 10934501 - -
          Affymetrix Rat Gene 1.0 ST Array 10767372 - -
          Affymetrix Rat Gene 1.0 ST Array 10937357 - -
          Affymetrix Rat Gene 1.0 ST Array 10899869 - -
          Affymetrix Rat Gene 1.0 ST Array 10878911 - -
          Affymetrix Rat Gene 1.0 ST Array 10899872 - -
          Affymetrix Rat Gene 1.1 ST Array 10899868 - -
          Affymetrix Rat Gene 1.1 ST Array 10751767 - -
          Affymetrix Rat Gene 1.1 ST Array 10934500 - -
          Affymetrix Rat Gene 1.1 ST Array 10878910 - -
          Affymetrix Rat Gene 1.1 ST Array 10767371 - -
          Affymetrix Rat Gene 1.1 ST Array 10937356 - -
          Rat Genome 230 2.0 Array 1367596_PM_at - -
          Rat Genome 230 2.0 Array 1367596_at - -
          Affymetrix HT Rat Focus Array 1367596_at - -
          Affymetrix HT Rat Focus Array 1367596_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700033131.rn.1 TC0700004044.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700033127.rn.1 TC0700004044.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700033129.rn.1 TC0700004044.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700033125.rn.1 TC0700004044.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700033126.rn.1 TC0700004044.rn.1 RTA TC
          Rat Expression Set 230 1367596_at - -
          Rat Expression Set 230 1367596_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10878910 - -
          Affymetrix Rat Gene 1.0 ST Array 10751767 - -
          Affymetrix Rat Gene 1.0 ST Array 10934500 - -
          Affymetrix Rat Gene 1.0 ST Array 10899868 - -
          Affymetrix Rat Gene 1.0 ST Array 10767371 - -
          Affymetrix Rat Gene 1.0 ST Array 10937356 - -
          Rat Genome U34 Set rc_AI014087_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22025978 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369103 - -
          RatRef-12 Expression BeadChip ILMN_50133 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0