Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00820764_g1
          See other SORD GE Assays ›
          Gene Symbol:
          Sord
          Gene Name:
          sorbitol dehydrogenase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 114176309 - 114207368 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00820764_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sord
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017052.2 NP_058748.2 6 - 7 697 65
          GenBank mRNA BC088398.1 - 6 - 7 697 65
          BC098919.1 - 6 - 7 644 65
          BC128707.1 - 6 - 7 673 65
          FQ234001.1 - 6 - 7 586 65
          X59037.1 - 6 - 7 637 65

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sord

          Gene Ontology Categories:

          Function(s) Process(es)

          sorbitol metabolic process
          sorbitol catabolic process
          response to osmotic stress
          response to xenobiotic stimulus
          response to hormone
          flagellated sperm motility
          response to nutrient levels
          fructose biosynthetic process
          response to cadmium ion
          response to copper ion
          L-xylitol catabolic process
          L-xylitol metabolic process
          L-iditol 2-dehydrogenase activity
          zinc ion binding
          identical protein binding
          NAD binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1000935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920325 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1772 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130144 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175233 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1000935 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1000935 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1000935 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1000935 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41450 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12130 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10839260 - -
          Affymetrix Rat Gene 1.0 ST Array 10839256 - -
          Affymetrix Rat Gene 1.0 ST Array 10932081 - -
          Affymetrix Rat Gene 1.0 ST Array 10839259 - -
          Affymetrix Rat Gene 1.0 ST Array 10839257 - -
          Affymetrix Rat Gene 1.0 ST Array 10839255 - -
          Affymetrix Rat Gene 1.0 ST Array 10839258 - -
          Affymetrix Rat Gene 1.1 ST Array 10932078 - -
          Affymetrix Rat Gene 1.1 ST Array 10839254 - -
          Rat Genome 230 2.0 Array 1369635_at - -
          Rat Genome 230 2.0 Array 1369635_PM_at - -
          Rat Genome 230 2.0 Array 1369636_PM_at - -
          Rat Genome 230 2.0 Array 1369636_at - -
          Affymetrix HT Rat Focus Array 1369635_PM_at - -
          Affymetrix HT Rat Focus Array 1369635_at - -
          Affymetrix HT Rat Focus Array 1369636_at - -
          Affymetrix HT Rat Focus Array 1369636_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300011082.rn.1 TC0300001587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300011086.rn.1 TC0300001587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300011081.rn.1 TC0300001587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300011087.rn.1 TC0300001587.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300011085.rn.1 TC0300001587.rn.1 RTA TC
          Rat Expression Set 230 1369635_PM_at - -
          Rat Expression Set 230 1369636_PM_at - -
          Rat Expression Set 230 1369635_at - -
          Rat Expression Set 230 1369636_at - -
          Affymetrix Rat Gene 1.0 ST Array 10839254 - -
          Rat Genome U34 Set rc_AI030175_s_at - -
          Rat Genome U34 Set X74593_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21852600 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366773 - -
          RatRef-12 Expression BeadChip ILMN_57551 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0