Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821099_g1
          See other RPL27 GE Assays ›
          Gene Symbol:
          Rpl27
          Gene Name:
          ribosomal protein L27
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 89352864 - 89356563 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821099_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpl27
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022514.1 NP_071959.1 2 - 3 265 119
          GenBank mRNA BC058474.1 - 3 - 4 295 119
          BC091566.1 - 3 - 4 263 119
          FQ210011.1 - 3 - 4 281 119
          FQ214023.1 - 3 - 4 285 119
          FQ217713.1 - 2 - 3 205 119
          FQ221292.1 - 3 - 4 290 119
          FQ221643.1 - 3 - 4 282 119
          FQ221684.1 - 3 - 4 283 119
          FQ221938.1 - 3 - 4 296 119
          FQ222018.1 - 3 - 4 285 119
          FQ222606.1 - 3 - 4 286 119
          FQ222991.1 - 3 - 4 287 119
          FQ223942.1 - 2 - 3 214 119
          FQ224664.1 - 3 - 4 283 119
          FQ228505.1 - 3 - 4 282 119
          FQ228603.1 - 3 - 4 283 119
          FQ228786.1 - 3 - 4 282 119
          X07424.1 - 2 - 3 265 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000004374.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpl27

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          RNA binding
          structural constituent of ribosome

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P088940 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P304257 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P110013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520520 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10333 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P088940 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P304257 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P110013 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P110013 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P304257 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P088940 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P110013 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P088940 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P304257 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P463378 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14014 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700143064 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20044 - -
          Agilent Rat cDNA Microarray (G4105A) 700143064 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10797764 - -
          Affymetrix Rat Gene 1.0 ST Array 10738414 - -
          Affymetrix Rat Gene 1.0 ST Array 10738412 - -
          Affymetrix Rat Gene 1.0 ST Array 10738410 - -
          Affymetrix Rat Gene 1.0 ST Array 10938580 - -
          Affymetrix Rat Gene 1.0 ST Array 10797765 - -
          Affymetrix Rat Gene 1.0 ST Array 10751238 - -
          Affymetrix Rat Gene 1.0 ST Array 10938581 - -
          Affymetrix Rat Gene 1.0 ST Array 10738411 - -
          Affymetrix Rat Gene 1.0 ST Array 10738413 - -
          Affymetrix Rat Gene 1.0 ST Array 10797764 - -
          Affymetrix Rat Gene 1.1 ST Array 10938579 - -
          Affymetrix Rat Gene 1.1 ST Array 10797763 - -
          Affymetrix Rat Gene 1.1 ST Array 10738409 - -
          Rat Genome 230 2.0 Array 1367561_at - -
          Rat Genome 230 2.0 Array 1367561_PM_at - -
          Affymetrix HT Rat Focus Array 1367561_PM_at - -
          Affymetrix HT Rat Focus Array 1367561_at - -
          Affymetrix Rat Clariom S Assay PSR1000040720.rn.1 TC1000004374.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000040719.rn.1 TC1000004374.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000040713.rn.1 TC1000004374.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000040725.rn.1 TC1000004374.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000040723.rn.1 TC1000004374.rn.1 RTA TC
          Rat Expression Set 230 1367561_at - -
          Rat Expression Set 230 1367561_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10938579 - -
          Affymetrix Rat Gene 1.0 ST Array 10751237 - -
          Affymetrix Rat Gene 1.0 ST Array 10797763 - -
          Affymetrix Rat Gene 1.0 ST Array 10738409 - -
          Rat Genome U34 Set rc_AI176589_g_at - -
          Rat Genome U34 Set rc_AI176589_at - -
          Rat Genome U34 Set rc_AI101165_at - -
          Rat Genome U34 Set rc_AI101165_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21797530 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367235 - -
          RatRef-12 Expression BeadChip ILMN_60946 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-ztd5s:80/100.66.134.6:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0