Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821130_g1
          See other RPS14 GE Assays ›
          Gene Symbol:
          Rps14 LOC100911847
          Gene Name:
          ribosomal protein S14
          40S ribosomal protein S14-like
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.18: 56042532 - 56047316 on Build Rnor_6.0
          Chromosome Location:
          Chr.5: 144083043 - 144083596 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn00821130_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rps14
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022672.1 NP_073163.1 3 - 4 347 88
          GenBank mRNA BC058472.1 - 2 - 3 354 88
          FQ210177.1 - 3 - 4 349 88
          FQ210421.1 - 3 - 4 349 88
          FQ210469.1 - 3 - 4 346 88
          FQ211405.1 - 3 - 4 347 88
          FQ211449.1 - 3 - 4 347 88
          FQ211623.1 - 3 - 4 349 88
          FQ212082.1 - 3 - 4 351 88
          FQ212093.1 - 3 - 4 347 88
          FQ212392.1 - 3 - 4 347 88
          FQ214636.1 - 3 - 4 349 88
          FQ217229.1 - 3 - 4 347 88
          FQ217605.1 - 3 - 4 347 88
          FQ218082.1 - 3 - 4 344 88
          FQ218760.1 - 3 - 4 349 88
          FQ219972.1 - 3 - 4 347 88
          FQ220057.1 - 3 - 4 347 88
          FQ220332.1 - 3 - 4 349 88
          FQ220676.1 - 3 - 4 351 88
          FQ220975.1 - 3 - 4 344 88
          FQ221057.1 - 3 - 4 347 88
          FQ221058.1 - 3 - 4 347 88
          FQ221066.1 - 3 - 4 347 88
          FQ221125.1 - 3 - 4 349 88
          FQ221205.1 - 3 - 4 347 88
          FQ221325.1 - 3 - 4 347 88
          FQ221335.1 - 3 - 4 347 88
          FQ221375.1 - 3 - 4 347 88
          FQ221450.1 - 3 - 4 347 88
          FQ221468.1 - 3 - 4 347 88
          FQ221581.1 - 3 - 4 346 88
          FQ221634.1 - 3 - 4 344 88
          FQ221754.1 - 3 - 4 343 88
          FQ221769.1 - 3 - 4 349 88
          FQ221835.1 - 3 - 4 349 88
          FQ221920.1 - 3 - 4 349 88
          FQ222066.1 - 3 - 4 344 88
          FQ222119.1 - 3 - 4 347 88
          FQ222124.1 - 3 - 4 347 88
          FQ222159.1 - 3 - 4 349 88
          FQ222163.1 - 3 - 4 348 88
          FQ222324.1 - 3 - 4 351 88
          FQ222370.1 - 3 - 4 347 88
          FQ222480.1 - 3 - 4 347 88
          FQ222552.1 - 3 - 4 346 88
          FQ222574.1 - 3 - 4 347 88
          FQ222575.1 - 2 - 3 347 88
          FQ222667.1 - 3 - 4 347 88
          FQ222697.1 - 3 - 4 349 88
          FQ222714.1 - 3 - 4 347 88
          FQ222747.1 - 3 - 4 351 88
          FQ222803.1 - 3 - 4 347 88
          FQ222840.1 - 3 - 4 350 88
          FQ222893.1 - 3 - 4 349 88
          FQ222979.1 - 3 - 4 349 88
          FQ222986.1 - 3 - 4 344 88
          FQ223007.1 - 3 - 4 347 88
          FQ223012.1 - 3 - 4 349 88
          FQ223069.1 - 3 - 4 349 88
          FQ223131.1 - 3 - 4 347 88
          FQ223204.1 - 3 - 4 349 88
          FQ223283.1 - 3 - 4 347 88
          FQ223454.1 - 3 - 4 348 88
          FQ223480.1 - 3 - 4 347 88
          FQ223519.1 - 3 - 4 346 88
          FQ223527.1 - 3 - 4 347 88
          FQ223570.1 - 3 - 4 347 88
          FQ224051.1 - 3 - 4 344 88
          FQ224105.1 - 3 - 4 349 88
          FQ224302.1 - 3 - 4 348 88
          FQ224493.1 - 3 - 4 349 88
          FQ225464.1 - 3 - 4 349 88
          FQ228360.1 - 3 - 4 332 88
          FQ228480.1 - 3 - 4 349 88
          FQ228486.1 - 3 - 4 347 88
          FQ228559.1 - 3 - 4 351 88
          FQ228608.1 - 3 - 4 347 88
          FQ228692.1 - 3 - 4 346 88
          FQ229006.1 - 3 - 4 353 88
          FQ229014.1 - 3 - 4 349 88
          FQ229055.1 - 3 - 4 349 88
          FQ229061.1 - 3 - 4 349 88
          FQ229229.1 - 3 - 4 349 88
          FQ229387.1 - 3 - 4 347 88
          FQ231306.1 - 490 88
          X15040.1 - 3 - 4 347 88
          Gene Symbol: LOC100911847
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003750030.3 XP_003750078.3 1 351 88

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rps14
          Click here to find siRNAs for Gene LOC100911847

          Gene Ontology Categories:

          Function(s) Process(es)

          ribosomal small subunit assembly
          negative regulation of transcription by RNA polymerase II
          translation
          regulation of translation
          erythrocyte differentiation
          maturation of SSU-rRNA
          RNA binding
          structural constituent of ribosome
          translation regulator activity
          mRNA 5'-UTR binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P334709 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P003090 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P132881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193097 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153671 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P334709 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P003090 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P334709 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P003090 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P334709 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P003090 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P334709 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7091 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10802010 - -
          Affymetrix Rat Gene 1.0 ST Array 10871998 - -
          Affymetrix Rat Gene 1.0 ST Array 10802008 - -
          Affymetrix Rat Gene 1.0 ST Array 10802012 - -
          Affymetrix Rat Gene 1.0 ST Array 10802009 - -
          Affymetrix Rat Gene 1.0 ST Array 10927282 - -
          Affymetrix Rat Gene 1.0 ST Array 10802011 - -
          Affymetrix Rat Gene 1.0 ST Array 10871999 - -
          Affymetrix Rat Gene 1.1 ST Array 10802007 - -
          Affymetrix Rat Gene 1.1 ST Array 10871997 - -
          Affymetrix Rat Gene 1.1 ST Array 10927281 - -
          Rat Genome 230 2.0 Array 1368211_PM_at - -
          Rat Genome 230 2.0 Array 1368211_at - -
          Affymetrix HT Rat Focus Array 1368211_at - -
          Affymetrix HT Rat Focus Array 1368211_PM_at - -
          Rat Expression Set 230 1368211_PM_at - -
          Rat Expression Set 230 1368211_at - -
          Affymetrix Rat Gene 1.0 ST Array 10871997 - -
          Affymetrix Rat Gene 1.0 ST Array 10927281 - -
          Affymetrix Rat Gene 1.0 ST Array 10802007 - -
          Rat Genome U34 Set rc_AA945806_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22323561 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371508 - -
          RatRef-12 Expression BeadChip ILMN_62109 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0