Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821133_g1
          See other H2AFZ GE Assays ›
          Gene Symbol:
          H2afz
          Gene Name:
          H2A histone family, member Z
          Gene Aliases:
          H2A.Z-1
          Chromosome Location:
          Chr.2: 243105224 - 243107333 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821133_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: H2afz
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022674.1 NP_073165.1 3 - 4 238 62
          GenBank mRNA BC060564.1 - 3 - 4 303 62
          BC086348.1 - 3 - 4 283 62
          FQ213837.1 - 3 - 4 297 62
          FQ220829.1 - 3 - 4 299 62
          FQ221791.1 - 3 - 4 297 62
          FQ222470.1 - 3 - 4 297 62
          FQ224994.1 - 3 - 4 300 62
          FQ227672.1 - 3 - 4 299 62
          FQ229641.1 - 3 - 4 299 62
          FQ229746.1 - 3 - 4 299 62
          FQ232362.1 - 3 - 4 299 62
          FQ233167.1 - 3 - 4 299 62
          FQ234498.1 - 3 - 4 299 62
          M37584.1 - 3 - 4 238 62
          X52316.1 - 3 - 4 209 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002276.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene H2afz

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular response to insulin stimulus
          positive regulation of transcription by RNA polymerase II
          cellular response to estradiol stimulus
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter sequence-specific DNA binding
          molecular_function
          DNA binding
          chromatin DNA binding
          nucleosomal DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P14682 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P009877 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P009877 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14682 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14682 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P009877 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P14682 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P009877 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14682 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10788859 - -
          Affymetrix Rat Gene 1.0 ST Array 10819324 - -
          Affymetrix Rat Gene 1.0 ST Array 10819326 - -
          Affymetrix Rat Gene 1.0 ST Array 10819328 - -
          Affymetrix Rat Gene 1.0 ST Array 10819327 - -
          Affymetrix Rat Gene 1.0 ST Array 10819323 - -
          Affymetrix Rat Gene 1.0 ST Array 10819325 - -
          Affymetrix Rat Gene 1.1 ST Array 10819322 - -
          Affymetrix Rat Gene 1.1 ST Array 10788858 - -
          Rat Genome 230 2.0 Array 1386861_at - -
          Rat Genome 230 2.0 Array 1386861_PM_at - -
          Affymetrix HT Rat Focus Array 1386861_at - -
          Affymetrix HT Rat Focus Array 1386861_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200015811.rn.1 TC0200002276.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200015814.rn.1 TC0200002276.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200015815.rn.1 TC0200002276.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200015812.rn.1 TC0200002276.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200015813.rn.1 TC0200002276.rn.1 RTA TC
          Rat Expression Set 230 1386861_at - -
          Rat Expression Set 230 1386861_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10788858 - -
          Affymetrix Rat Gene 1.0 ST Array 10819322 - -
          Rat Genome U34 Set M37584_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21396182 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351035 - -
          RatRef-12 Expression BeadChip ILMN_56622 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0