Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821258_g1
          See other RPL13 GE Assays ›
          Gene Symbol:
          Rpl13 LOC100359922
          Gene Name:
          ribosomal protein L13
          60S ribosomal protein L13-like
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.19: 55917489 - 55920040 on Build Rnor_6.0
          Chromosome Location:
          Chr.8: 6749722 - 6750454 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn00821258_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpl13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031101.1 NP_112363.1 5 - 6 515 68
          GenBank mRNA BC058143.1 - 5 - 6 518 68
          BC086577.1 - 3 - 4 739 68
          FQ222628.1 - 5 - 6 513 68
          FQ222810.1 - 5 - 6 513 68
          FQ223361.1 - 5 - 6 534 68
          FQ224984.1 - 5 - 6 511 68
          FQ225827.1 - 3 - 4 281 68
          FQ226346.1 - 5 - 6 512 68
          FQ228254.1 - 5 - 6 511 68
          X78327.1 - 5 - 6 515 68
          Gene Symbol: LOC100359922
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008765903.1 XP_008764125.1 1 512 68

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpl13
          Click here to find siRNAs for Gene LOC100359922

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          RNA binding
          structural constituent of ribosome

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P029673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325034 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P029673 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P029673 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P029673 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P138057 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P130780 - -
          Agilent Rat cDNA Microarray (G4105A) 701232278 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10914850 - -
          Affymetrix Rat Gene 1.0 ST Array 10914852 - -
          Affymetrix Rat Gene 1.0 ST Array 10777033 - -
          Affymetrix Rat Gene 1.0 ST Array 10808597 - -
          Affymetrix Rat Gene 1.0 ST Array 10808596 - -
          Affymetrix Rat Gene 1.0 ST Array 10808599 - -
          Affymetrix Rat Gene 1.0 ST Array 10808600 - -
          Affymetrix Rat Gene 1.0 ST Array 10774103 - -
          Affymetrix Rat Gene 1.0 ST Array 10851387 - -
          Affymetrix Rat Gene 1.0 ST Array 10733689 - -
          Affymetrix Rat Gene 1.0 ST Array 10883460 - -
          Affymetrix Rat Gene 1.0 ST Array 10914851 - -
          Affymetrix Rat Gene 1.0 ST Array 10808598 - -
          Affymetrix Rat Gene 1.1 ST Array 10851386 - -
          Affymetrix Rat Gene 1.1 ST Array 10883459 - -
          Affymetrix Rat Gene 1.1 ST Array 10777032 - -
          Affymetrix Rat Gene 1.1 ST Array 10914849 - -
          Affymetrix Rat Gene 1.1 ST Array 10774102 - -
          Affymetrix Rat Gene 1.1 ST Array 10733688 - -
          Affymetrix Rat Gene 1.1 ST Array 10808595 - -
          Rat Genome 230 2.0 Array 1386858_at - -
          Rat Genome 230 2.0 Array 1386858_PM_at - -
          Affymetrix HT Rat Focus Array 1386858_at - -
          Affymetrix HT Rat Focus Array 1386858_PM_at - -
          Rat Expression Set 230 1386858_at - -
          Rat Expression Set 230 1386858_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10777032 - -
          Affymetrix Rat Gene 1.0 ST Array 10914849 - -
          Affymetrix Rat Gene 1.0 ST Array 10733688 - -
          Affymetrix Rat Gene 1.0 ST Array 10851386 - -
          Affymetrix Rat Gene 1.0 ST Array 10808595 - -
          Affymetrix Rat Gene 1.0 ST Array 10774102 - -
          Rat Genome U34 Set X78327_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354937 - -
          RatRef-12 Expression BeadChip ILMN_58529 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7ffdcc7b59-b9k6r:80/100.66.134.75:80.
          git-commit: c6f4609997b4833aba2fd3ad806738f0433cd95c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc