Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821295_g1
          See other RPS27A GE Assays ›
          Gene Symbol:
          Rps27a LOC100912032 LOC681260 Rps27l3
          Gene Name:
          ribosomal protein S27a
          ubiquitin-40S ribosomal protein S27a-like
          similar to ribosomal protein S27a
          ribosomal protein S27-like 3
          Gene Aliases:
          Uba52, Ubb
          -
          -
          Gm8430, RGD1564290
          Chromosome Location:
          Chr.5: 138628482 - 138629026 on Build Rnor_6.0
          Chromosome Location:
          Chr.14: 113966573 - 113968469 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 255572806 - 255573402 on Build Rnor_6.0
          Chromosome Location:
          Chr.18: 25394170 - 25394839 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          Assay ID Rn00821295_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rps27a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031113.1 NP_112375.1 1 361 64
          Gene Symbol: LOC100912032
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003751421.3 XP_003751469.1 5 - 6 425 64
          Gene Symbol: LOC681260
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008760378.1 XP_008758600.1 2 344 64
          Gene Symbol: Rps27l3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008772051.1 XP_008770273.1 2 343 64
          XM_574121.5 XP_574121.1 2 341 64

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rps27a
          Click here to find siRNAs for Gene LOC100912032
          Click here to find siRNAs for Gene LOC681260
          Click here to find siRNAs for Gene Rps27l3

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          RNA binding
          structural constituent of ribosome
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P367214 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P146096 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P160412 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P293187 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P144208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782738 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68915 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P146096 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P160412 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P293187 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P144208 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P367214 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P160412 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P146096 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P367214 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P144208 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P293187 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P293187 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P367214 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P160412 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P144208 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P146096 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P561580 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P293187 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700174880 - -
          Agilent Rat cDNA Microarray (G4105A) 700174880 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10803677 - -
          Affymetrix Rat Gene 1.0 ST Array 10803678 - -
          Affymetrix Rat Gene 1.0 ST Array 10778855 - -
          Affymetrix Rat Gene 1.0 ST Array 10778856 - -
          Affymetrix Rat Gene 1.0 ST Array 10778854 - -
          Affymetrix Rat Gene 1.0 ST Array 10778858 - -
          Affymetrix Rat Gene 1.0 ST Array 10879393 - -
          Affymetrix Rat Gene 1.0 ST Array 10778857 - -
          Affymetrix Rat Gene 1.1 ST Array 10803676 - -
          Affymetrix Rat Gene 1.1 ST Array 10778853 - -
          Affymetrix Rat Gene 1.1 ST Array 10879392 - -
          Rat Genome 230 2.0 Array 1367685_at - -
          Rat Genome 230 2.0 Array 1367685_PM_at - -
          Rat Genome 230 2.0 Array 1398617_at - -
          Rat Genome 230 2.0 Array 1398617_PM_at - -
          Affymetrix HT Rat Focus Array 1367685_at - -
          Affymetrix HT Rat Focus Array 1367685_PM_at - -
          Rat Expression Set 230 1367685_at - -
          Rat Expression Set 230 1367685_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10879392 - -
          Affymetrix Rat Gene 1.0 ST Array 10803676 - -
          Affymetrix Rat Gene 1.0 ST Array 10803676 - -
          Affymetrix Rat Gene 1.0 ST Array 10879392 - -
          Affymetrix Rat Gene 1.0 ST Array 10879392 - -
          Affymetrix Rat Gene 1.0 ST Array 10778853 - -
          Rat Genome U34 Set rc_AA891729_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21681836 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362582 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365412 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372204 - -
          RatRef-12 Expression BeadChip ILMN_67113 - -
          RatRef-12 Expression BeadChip ILMN_68667 - -
          RatRef-12 Expression BeadChip ILMN_52062 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-998b77544-nf2nv:80/100.66.134.75:80.
          git-commit: efd2b0e08ab99238c7874a09efb06372d741ac0a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc