Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821391_g1
          See other SCD2 GE Assays ›
          Gene Symbol:
          Scd2
          Gene Name:
          stearoyl-Coenzyme A desaturase 2
          Gene Aliases:
          Scd
          Chromosome Location:
          Chr.1: 264059374 - 264072433 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821391_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Scd2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031841.1 NP_114029.1 1 - 2 369 67
          GenBank mRNA AB032243.1 - 1 - 2 369 67
          BC061737.1 - 1 - 2 311 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003679.rn.1

          Additional Information:

          This assay is in the 5'UTR region of the AB032243.1, BC061737.1, NM_031841.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Scd2

          Gene Ontology Categories:

          Function(s) Process(es)

          fatty acid biosynthetic process
          myelination
          stearoyl-CoA 9-desaturase activity
          iron ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P813567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741160 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94594 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150580 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P813567 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P813567 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P813567 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P198396 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P813567 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P194152 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102417 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92884 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10715550 - -
          Affymetrix Rat Gene 1.0 ST Array 10715551 - -
          Affymetrix Rat Gene 1.0 ST Array 10715548 - -
          Affymetrix Rat Gene 1.0 ST Array 10715549 - -
          Affymetrix Rat Gene 1.0 ST Array 10715547 - -
          Affymetrix Rat Gene 1.1 ST Array 10715546 - -
          Rat Genome 230 2.0 Array 1367668_a_at - -
          Rat Genome 230 2.0 Array 1388253_at - -
          Rat Genome 230 2.0 Array 1386889_at - -
          Rat Genome 230 2.0 Array 1398016_at - -
          Rat Genome 230 2.0 Array 1386889_PM_at - -
          Rat Genome 230 2.0 Array 1367668_PM_a_at - -
          Rat Genome 230 2.0 Array 1388253_PM_at - -
          Rat Genome 230 2.0 Array 1398016_PM_at - -
          Affymetrix HT Rat Focus Array 1398016_at - -
          Affymetrix HT Rat Focus Array 1367668_PM_a_at - -
          Affymetrix HT Rat Focus Array 1386889_PM_at - -
          Affymetrix HT Rat Focus Array 1398016_PM_at - -
          Affymetrix HT Rat Focus Array 1386889_at - -
          Affymetrix HT Rat Focus Array 1388253_at - -
          Affymetrix HT Rat Focus Array 1388253_PM_at - -
          Affymetrix HT Rat Focus Array 1367668_a_at - -
          Affymetrix Rat Clariom S Assay PSR0100029898.rn.1 TC0100003679.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100029903.rn.1 TC0100003679.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100029897.rn.1 TC0100003679.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100029901.rn.1 TC0100003679.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100029904.rn.1 TC0100003679.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100029899.rn.1 TC0100003679.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100029900.rn.1 TC0100003679.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100029905.rn.1 TC0100003679.rn.1 RTA TC
          Rat Expression Set 230 1367668_a_at - -
          Rat Expression Set 230 1388253_PM_at - -
          Rat Expression Set 230 1388253_at - -
          Rat Expression Set 230 1386889_at - -
          Rat Expression Set 230 1386889_PM_at - -
          Rat Expression Set 230 1367668_PM_a_at - -
          Rat Expression Set 230 1398016_at - -
          Rat Expression Set 230 1398016_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10715546 - -
          Rat Genome U34 Set AF036761_at - -
          Rat Genome U34 Set AF036761_g_at - -
          Rat Genome U34 Set S75730_at - -
          Rat Genome U34 Set M15114_at - -
          Rat Genome U34 Set U67995_s_at - -
          Rat Genome U34 Set rc_AA875269_at - -
          Rat Genome U34 Set M15114_g_at - -
          Rat Genome U34 Set rc_AI011483_at - -
          Rat Genome U34 Set rc_AI228830_s_at - -
          Rat Genome U34 Set M15114_at - -
          Rat Genome U34 Set M15114_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20786621 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359586 - -
          RatRef-12 Expression BeadChip ILMN_52257 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0