Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821494_g1
          See other BANF1 GE Assays ›
          Gene Symbol:
          Banf1
          Gene Name:
          barrier to autointegration factor 1
          Gene Aliases:
          Baf, Bcrp1, L2bp1, L2bp1/Baf
          Chromosome Location:
          Chr.1: 220744195 - 220746224 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821494_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Banf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053631.3 NP_446083.1 2 - 3 217 95
          XM_006230630.2 XP_006230692.1 2 - 3 230 95
          XM_006230631.1 XP_006230693.1 2 - 3 254 95
          XM_006230632.2 XP_006230694.1 2 - 3 227 95
          GenBank mRNA AB024333.1 - 2 - 3 212 95
          BC084726.1 - 2 - 3 188 95
          CB766065.1 - 2 - 3 217 95
          FQ224698.1 - 2 - 3 259 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007110.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Banf1

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA integration
          regulation of viral process
          DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P093412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293294 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P093412 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P093412 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P093412 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P671364 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700372473 - -
          Agilent Rat cDNA Microarray (G4105A) 700372473 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10727813 - -
          Affymetrix Rat Gene 1.0 ST Array 10727814 - -
          Affymetrix Rat Gene 1.1 ST Array 10727812 - -
          Rat Genome 230 2.0 Array 1367820_at - -
          Rat Genome 230 2.0 Array 1367820_PM_at - -
          Affymetrix HT Rat Focus Array 1367820_at - -
          Affymetrix HT Rat Focus Array 1367820_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100056793.rn.1 TC0100007110.rn.1 RTA TC
          Rat Expression Set 230 1367820_at - -
          Rat Expression Set 230 1367820_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10727812 - -
          Rat Genome U34 Set AA848386_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22179362 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364653 - -
          RatRef-12 Expression BeadChip ILMN_58077 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0