Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821577_g1
          See other RGCC GE Assays ›
          Gene Symbol:
          Rgcc
          Gene Name:
          regulator of cell cycle
          Gene Aliases:
          Rgc32
          Chromosome Location:
          Chr.15: 61551861 - 61564695 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821577_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rgcc
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_054008.1 NP_446460.1 4 - 5 493 100
          GenBank mRNA AF036548.1 - 4 - 5 493 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500002140.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rgcc

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cyclin-dependent protein serine/threonine kinase activity
          negative regulation of exit from mitosis
          negative regulation of cytokine production
          positive regulation of cytokine production
          negative regulation of endothelial cell proliferation
          positive regulation of extracellular matrix constituent secretion
          complement activation
          negative regulation of cell population proliferation
          positive regulation of gene expression
          positive regulation of epithelial to mesenchymal transition
          negative regulation of angiogenesis
          positive regulation of collagen biosynthetic process
          negative regulation of blood vessel endothelial cell migration
          positive regulation of mitotic nuclear division
          positive regulation of transcription by RNA polymerase II
          positive regulation of DNA-binding transcription factor activity
          positive regulation of stress fiber assembly
          regulation of cell cycle
          cellular response to hypoxia
          fibroblast activation
          negative regulation of fibroblast growth factor production
          positive regulation of extracellular matrix assembly
          negative regulation of mitotic cell cycle phase transition
          negative regulation of cell-cell adhesion mediated by cadherin
          positive regulation of endothelial cell apoptotic process
          positive regulation of DNA biosynthetic process
          protein kinase binding
          protein kinase activator activity
          R-SMAD binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P150904 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P102942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89931 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P102942 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P150904 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P102942 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P150904 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P150904 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P102942 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P493270 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700294711 - -
          Agilent Rat cDNA Microarray (G4105A) 700294711 - -
          Array Name Probe IDs Targets Target Type
          Rat Genome 230 2.0 Array 1368080_PM_at - -
          Rat Genome 230 2.0 Array 1368080_at - -
          Rat Genome 230 2.0 Array 1389972_at - -
          Rat Genome 230 2.0 Array 1389972_PM_at - -
          Affymetrix HT Rat Focus Array 1368080_PM_at - -
          Affymetrix HT Rat Focus Array 1368080_at - -
          Affymetrix Rat Clariom S Assay PSR1500014714.rn.1 TC1500002140.rn.1 RTA TC
          Rat Expression Set 230 1368080_at - -
          Rat Expression Set 230 1368080_PM_at - -
          Rat Expression Set 230 1389972_PM_at - -
          Rat Expression Set 230 1389972_at - -
          Rat Genome U34 Set AF036548_g_at - -
          Rat Genome U34 Set AF036548_at - -
          Rat Genome U34 Set rc_AA859967_at - -
          Rat Genome U34 Set rc_AI178983_at - -
          Rat Genome U34 Set rc_AI175914_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22120833 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1530263 - -
          RatRef-12 Expression BeadChip ILMN_161402 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0