Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821655_g1
          See other HIGD1A GE Assays ›
          Gene Symbol:
          Higd1a
          Gene Name:
          HIG1 hypoxia inducible domain family, member 1A
          Gene Aliases:
          Hig1
          Chromosome Location:
          Chr.8: 130482492 - 130491888 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821655_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Higd1a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_080902.4 NP_543178.2 3 - 4 317 73
          GenBank mRNA AY062253.3 - 3 - 4 376 73
          BC059118.1 - 3 - 4 321 73

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Higd1a

          Gene Ontology Categories:

          Function(s)

          cellular response to glucose starvation
          negative regulation of apoptotic process
          cellular response to hypoxia
          negative regulation of release of cytochrome c from mitochondria

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P306507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864657 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82316 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P306507 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P306507 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P306507 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13096 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P306507 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42217 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10914416 - -
          Affymetrix Rat Gene 1.0 ST Array 10914417 - -
          Affymetrix Rat Gene 1.0 ST Array 10914418 - -
          Affymetrix Rat Gene 1.1 ST Array 10914415 - -
          Rat Genome 230 2.0 Array 1370062_at - -
          Rat Genome 230 2.0 Array 1370062_PM_at - -
          Rat Genome 230 2.0 Array 1374105_at - -
          Rat Genome 230 2.0 Array 1374105_PM_at - -
          Affymetrix HT Rat Focus Array 1374105_PM_at - -
          Affymetrix HT Rat Focus Array 1374105_at - -
          Affymetrix HT Rat Focus Array 1370062_at - -
          Affymetrix HT Rat Focus Array 1370062_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800029246.rn.1 TC0800003590.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800029243.rn.1 TC0800003590.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800029244.rn.1 TC0800003590.rn.1 RTA TC
          Rat Expression Set 230 1370062_PM_at - -
          Rat Expression Set 230 1374105_at - -
          Rat Expression Set 230 1370062_at - -
          Rat Expression Set 230 1374105_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10914415 - -
          Rat Genome U34 Set rc_AA891422_at - -
          Rat Genome U34 Set rc_H31665_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22171111 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650956 - -
          RatRef-12 Expression BeadChip ILMN_50513 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix