Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821687_g1
          See other GNB2L1 GE Assays ›
          Gene Symbol:
          Gnb2l1
          Gene Name:
          guanine nucleotide binding protein (G protein), beta polypeptide 2 like 1
          Gene Aliases:
          RACK1
          Chromosome Location:
          Chr.10: 34149717 - 34155198 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821687_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gnb2l1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_130734.2 NP_570090.2 7 - 8 1021 63
          GenBank mRNA BC063809.1 - 7 - 8 996 63
          FQ212638.1 - 7 - 8 992 63
          FQ215527.1 - 7 - 8 993 63
          FQ215812.1 - 7 - 8 992 63
          FQ216167.1 - 7 - 8 992 63
          FQ216212.1 - 7 - 8 992 63
          FQ219492.1 - 7 - 8 993 63
          FQ219729.1 - 7 - 8 992 63
          FQ219878.1 - 7 - 8 990 63
          FQ219900.1 - 7 - 8 992 63
          FQ220124.1 - 7 - 8 992 63
          FQ220151.1 - 7 - 8 992 63
          FQ220277.1 - 7 - 8 992 63
          FQ220314.1 - 7 - 8 992 63
          FQ220391.1 - 7 - 8 992 63
          FQ220428.1 - 7 - 8 996 63
          FQ220430.1 - 7 - 8 991 63
          FQ220494.1 - 7 - 8 992 63
          FQ220598.1 - 7 - 8 996 63
          FQ220697.1 - 7 - 8 996 63
          FQ220819.1 - 7 - 8 993 63
          FQ220976.1 - 7 - 8 999 63
          FQ221286.1 - 7 - 8 996 63
          FQ222502.1 - 7 - 8 992 63
          FQ223855.1 - 7 - 8 991 63
          FQ224952.1 - 7 - 8 992 63
          FQ225047.1 - 7 - 8 996 63
          FQ225312.1 - 7 - 8 991 63
          FQ225511.1 - 7 - 8 992 63
          FQ225843.1 - 7 - 8 1022 63
          FQ226802.1 - 7 - 8 992 63
          FQ228007.1 - 7 - 8 996 63
          FQ229157.1 - 7 - 8 63
          FQ229247.1 - 7 - 8 992 63
          FQ229458.1 - 7 - 8 989 63
          FQ229569.1 - 7 - 8 993 63
          FQ229633.1 - 7 - 8 992 63
          FQ229689.1 - 7 - 8 992 63
          FQ229758.1 - 7 - 8 992 63
          FQ229908.1 - 7 - 8 993 63
          FQ229923.1 - 7 - 8 991 63
          FQ230003.1 - 7 - 8 990 63
          FQ230018.1 - 7 - 8 1000 63
          FQ230117.1 - 7 - 8 992 63
          FQ230328.1 - 7 - 8 994 63
          FQ233024.1 - 7 - 8 992 63
          FQ234000.1 - 7 - 8 998 63
          FQ234673.1 - 7 - 8 994 63
          FQ235255.1 - 7 - 8 992 63
          U03390.1 - 7 - 8 984 63

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gnb2l1

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          positive regulation of protein phosphorylation
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          protein kinase C-activating G protein-coupled receptor signaling pathway
          gastrulation
          negative regulation of gene expression
          negative regulation of translation
          negative regulation of Wnt signaling pathway
          negative regulation of cell growth
          positive regulation of cell migration
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          positive regulation of protein homooligomerization
          regulation of protein localization
          positive regulation of apoptotic process
          positive regulation of GTPase activity
          rhythmic process
          negative regulation of phagocytosis
          regulation of cell division
          positive regulation of cyclic-nucleotide phosphodiesterase activity
          regulation of cell cycle
          negative regulation of protein kinase B signaling
          positive regulation of mitochondrial depolarization
          negative regulation of protein tyrosine kinase activity
          regulation of protein localization to plasma membrane
          negative regulation of hydrogen peroxide-induced neuron death
          regulation of establishment of cell polarity
          positive regulation of gastrulation
          positive regulation of intrinsic apoptotic signaling pathway
          RNA binding
          protein kinase C binding
          ion channel inhibitor activity
          cysteine-type endopeptidase activator activity involved in apoptotic process
          enzyme binding
          protein phosphatase binding
          protein tyrosine kinase inhibitor activity
          receptor tyrosine kinase binding
          SH2 domain binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P447368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892691 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88750 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P447368 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P447368 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P447368 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P447368 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701882554 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700147481 - -
          Agilent Rat cDNA Microarray (G4105A) 701882554 - -
          Agilent Rat cDNA Microarray (G4105A) 700147481 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10733044 - -
          Affymetrix Rat Gene 1.0 ST Array 10733040 - -
          Affymetrix Rat Gene 1.0 ST Array 10733042 - -
          Affymetrix Rat Gene 1.0 ST Array 10733043 - -
          Affymetrix Rat Gene 1.0 ST Array 10733041 - -
          Affymetrix Rat Gene 1.1 ST Array 10733039 - -
          Rat Genome 230 2.0 Array 1367618_PM_a_at - -
          Rat Genome 230 2.0 Array 1367618_a_at - -
          Affymetrix HT Rat Focus Array 1367618_a_at - -
          Affymetrix HT Rat Focus Array 1367618_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR1000003906.rn.1 TC1000000466.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000003907.rn.1 TC1000000466.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000003909.rn.1 TC1000000466.rn.1 RTA TC
          Rat Expression Set 230 1367618_a_at - -
          Rat Expression Set 230 1367618_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10733039 - -
          Rat Genome U34 Set U03390_at - -
          Rat Genome U34 Set rc_AI171319_s_at - -
          Rat Genome U34 Set rc_AI171319_s_at - -
          Rat Genome U34 Set U03390_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22318653 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374713 - -
          RatRef-12 Expression BeadChip ILMN_55597 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0