Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821752_g1
          See other GZMB GE Assays ›
          Gene Symbol:
          Gzmb Gzmbl2
          Gene Name:
          granzyme B (granzyme 2, cytotoxic T-lymphocyte-associated serine esterase 1)
          Granzyme B-like 2
          Gene Aliases:
          RNKP-1
          -
          Chromosome Location:
          Chr.15: 35413862 - 35417293 on Build Rnor_6.0
          Chromosome Location:
          Chr.15: 36469315 - 36472395 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn00821752_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gzmb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138517.3 NP_612526.2 3 - 4 407 87
          GenBank mRNA BC127475.1 - 3 - 4 387 87
          FQ221957.1 - 3 - 4 429 87
          FQ228927.1 - 3 - 4 429 87
          Gene Symbol: Gzmbl2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_002728257.4 XP_002728303.2 3 - 4 475 87

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gzmb
          Click here to find siRNAs for Gene Gzmbl2

          Gene Ontology Categories:

          Function(s) Process(es)

          T cell mediated cytotoxicity
          proteolysis
          immune response
          granzyme-mediated apoptotic signaling pathway
          positive regulation of cell death
          protein processing
          cytolysis
          natural killer cell mediated cytotoxicity
          positive regulation of necroptotic process
          serine-type endopeptidase activity
          protein binding
          serine-type peptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P062980 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P153427 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P004796 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P16029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861586 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50921 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97619 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128121 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5474 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178794 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163566 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82827 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P062980 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P004796 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P004796 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P062980 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P004796 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P062980 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P16029 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P109503 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29955 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64702 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33273 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10424 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32580 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10784058 - -
          Affymetrix Rat Gene 1.0 ST Array 10784066 - -
          Affymetrix Rat Gene 1.0 ST Array 10784053 - -
          Affymetrix Rat Gene 1.0 ST Array 10784057 - -
          Affymetrix Rat Gene 1.0 ST Array 10784055 - -
          Affymetrix Rat Gene 1.0 ST Array 10784069 - -
          Affymetrix Rat Gene 1.0 ST Array 10784051 - -
          Affymetrix Rat Gene 1.0 ST Array 10784052 - -
          Affymetrix Rat Gene 1.0 ST Array 10784068 - -
          Affymetrix Rat Gene 1.0 ST Array 10784050 - -
          Affymetrix Rat Gene 1.0 ST Array 10875987 - -
          Affymetrix Rat Gene 1.0 ST Array 10784056 - -
          Affymetrix Rat Gene 1.0 ST Array 10784067 - -
          Affymetrix Rat Gene 1.1 ST Array 10784049 - -
          Affymetrix Rat Gene 1.1 ST Array 10784054 - -
          Affymetrix Rat Gene 1.1 ST Array 10875985 - -
          Affymetrix Rat Gene 1.1 ST Array 10784065 - -
          Rat Genome 230 2.0 Array 1370628_PM_at - -
          Rat Genome 230 2.0 Array 1371126_x_at - -
          Rat Genome 230 2.0 Array 1371126_PM_x_at - -
          Rat Genome 230 2.0 Array 1379794_at - -
          Rat Genome 230 2.0 Array 1370628_at - -
          Rat Genome 230 2.0 Array 1379794_PM_at - -
          Affymetrix HT Rat Focus Array 1371126_PM_x_at - -
          Affymetrix HT Rat Focus Array 1379794_at - -
          Affymetrix HT Rat Focus Array 1371126_x_at - -
          Affymetrix HT Rat Focus Array 1379794_PM_at - -
          Affymetrix HT Rat Focus Array 1370628_PM_at - -
          Affymetrix HT Rat Focus Array 1370628_at - -
          Affymetrix Rat Clariom S Assay PSR1500011539.rn.1 TC1500001745.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500011535.rn.1 TC1500001745.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500011537.rn.1 TC1500001745.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500011536.rn.1 TC1500001745.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500011538.rn.1 TC1500001745.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500011540.rn.1 TC1500001745.rn.1 RTA TC
          Rat Expression Set 230 1371126_PM_x_at - -
          Rat Expression Set 230 1371126_x_at - -
          Rat Expression Set 230 1370628_PM_at - -
          Rat Expression Set 230 1370628_at - -
          Rat Expression Set 230 1379794_at - -
          Rat Expression Set 230 1379794_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10784049 - -
          Affymetrix Rat Gene 1.0 ST Array 10784065 - -
          Affymetrix Rat Gene 1.0 ST Array 10784054 - -
          Rat Genome U34 Set X76996_f_at - -
          Rat Genome U34 Set M34097_at - -
          Rat Genome U34 Set X66693_f_at - -
          Rat Genome U34 Set rc_AA944850_s_at - -
          Rat Genome U34 Set rc_AI029386_i_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21539290 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358992 - -
          RatRef-12 Expression BeadChip ILMN_49667 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0