Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00822023_g1
          See other HSP90AA1 GE Assays ›
          Gene Symbol:
          Hsp90aa1 LOC103692716
          Gene Name:
          heat shock protein 90, alpha (cytosolic), class A member 1
          heat shock protein HSP 90-alpha
          Gene Aliases:
          Hsp86, Hsp90, Hspca
          -
          Chromosome Location:
          Chr.6: 135107262 - 135112793 on Build Rnor_6.0
          Chromosome Location:
          Chr.6: 135044833 - 135050421 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn00822023_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hsp90aa1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175761.2 NP_786937.1 10 - 11 2142 87
          GenBank mRNA AJ428213.1 - 10 - 11 2142 87
          BC072489.1 - 10 - 11 2124 7105514 87
          BC085120.1 - 10 - 11 2124 7114546 87
          FQ212905.1 - 4 - 5 772 87
          FQ213418.1 - 1 - 2 69 87
          FQ226694.1 - 1 - 2 69 87
          FQ226837.1 - 1 - 2 87
          FQ233994.1 - 1 - 2 69 87
          Gene Symbol: LOC103692716
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008764969.1 XP_008763191.1 2194 87

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hsp90aa1
          Click here to find siRNAs for Gene LOC103692716

          Panther Classification:

          Molecular Function -

          Hsp90 family chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          neuron migration
          skeletal muscle contraction
          protein folding
          response to heat
          response to salt stress
          positive regulation of lamellipodium assembly
          cardiac muscle cell apoptotic process
          positive regulation of protein import into nucleus, translocation
          response to estrogen
          protein import into mitochondrial outer membrane
          positive regulation of nitric oxide biosynthetic process
          positive regulation of cell size
          regulation of catalytic activity
          chaperone-mediated protein complex assembly
          positive regulation of cardiac muscle contraction
          UTP binding
          CTP binding
          mRNA binding
          protein binding
          ATP binding
          GTP binding
          ATPase activity
          sulfonylurea receptor binding
          protein kinase binding
          protein phosphatase binding
          MHC class II protein complex binding
          nitric-oxide synthase regulator activity
          TPR domain binding
          dATP binding
          identical protein binding
          protein homodimerization activity
          ion channel binding
          poly(A) RNA binding
          Rho GDP-dissociation inhibitor binding
          unfolded protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P299870 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P119852 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P023633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633917 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46439 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119852 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P023633 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P299870 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P023633 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P299870 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119852 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P119852 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P023633 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P299870 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P261874 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P135339 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P299870 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P114308 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71364 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10892191 - -
          Affymetrix Rat Gene 1.0 ST Array 10892186 - -
          Affymetrix Rat Gene 1.0 ST Array 10892195 - -
          Affymetrix Rat Gene 1.0 ST Array 10892192 - -
          Affymetrix Rat Gene 1.0 ST Array 10892194 - -
          Affymetrix Rat Gene 1.0 ST Array 10892187 - -
          Affymetrix Rat Gene 1.0 ST Array 10892190 - -
          Affymetrix Rat Gene 1.0 ST Array 10892196 - -
          Affymetrix Rat Gene 1.0 ST Array 10892185 - -
          Affymetrix Rat Gene 1.0 ST Array 10892189 - -
          Affymetrix Rat Gene 1.0 ST Array 10892188 - -
          Affymetrix Rat Gene 1.0 ST Array 10892193 - -
          Affymetrix Rat Gene 1.1 ST Array 10892184 - -
          Rat Genome 230 2.0 Array 1372701_PM_at - -
          Rat Genome 230 2.0 Array 1372701_at - -
          Rat Genome 230 2.0 Array 1388850_at - -
          Rat Genome 230 2.0 Array 1388850_PM_at - -
          Rat Genome 230 2.0 Array 1385541_at - -
          Rat Genome 230 2.0 Array 1385541_PM_at - -
          Affymetrix HT Rat Focus Array 1388850_PM_at - -
          Affymetrix HT Rat Focus Array 1372701_PM_at - -
          Affymetrix HT Rat Focus Array 1372701_at - -
          Affymetrix HT Rat Focus Array 1388850_at - -
          Affymetrix HT Rat Focus Array 1385541_PM_at - -
          Affymetrix HT Rat Focus Array 1385541_at - -
          Affymetrix Rat Clariom S Assay PSR0600022578.rn.1 TC0600003193.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600022576.rn.1 TC0600003193.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600022583.rn.1 TC0600003193.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600022590.rn.1 TC0600003193.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600022581.rn.1 TC0600003193.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600022585.rn.1 TC0600003193.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600022586.rn.1 TC0600003193.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600022588.rn.1 TC0600003193.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600022589.rn.1 TC0600003193.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600022584.rn.1 TC0600003193.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600022587.rn.1 TC0600003193.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600022577.rn.1 TC0600003193.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600022594.rn.1 TC0600003193.rn.1 RTA TC
          Rat Expression Set 230 1385541_at - -
          Rat Expression Set 230 1385541_PM_at - -
          Rat Expression Set 230 1388850_PM_at - -
          Rat Expression Set 230 1372701_PM_at - -
          Rat Expression Set 230 1388850_at - -
          Rat Expression Set 230 1372701_at - -
          Affymetrix Rat Gene 1.0 ST Array 10892184 - -
          Rat Genome U34 Set rc_AI176546_at - -
          Rat Genome U34 Set rc_AA944397_at - -
          Rat Genome U34 Set rc_AA819776_f_at - -
          Rat Genome U34 Set rc_AA819776_f_at - -
          Rat Genome U34 Set rc_AA944397_at - -
          Rat Genome U34 Set rc_AI176546_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20844189 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358144 - -
          RatRef-12 Expression BeadChip ILMN_57186 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0