Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00824628_m1
          See other PTGDRL GE Assays ›
          Gene Symbol:
          Ptgdrl Ptgdr
          Gene Name:
          prostaglandin D2 receptor
          prostaglandin D2 receptor-like
          Gene Aliases:
          Ptgdr, Ptgdr2
          RGD1565108
          Chromosome Location:
          Chr.15: 19195606 - 19196508 on Build Rnor_6.0
          Chromosome Location:
          Chr.15: 18089077 - 18095979 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn00824628_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ptgdrl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022241.1 NP_071577.1 901 73
          GenBank mRNA U92289.1 - 901 73
          Gene Symbol: Ptgdr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001135164.1 NP_001128636.1 1 - 2 841 73
          GenBank mRNA AF120101.1 - 1 - 2 841 73
          FQ222068.1 - 943 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001403.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Rattus norvegicus Rat Inflammation Inventoried Rat_Inflammation Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ptgdrl
          Click here to find siRNAs for Gene Ptgdr

          Gene Ontology Categories:

          Function(s) Process(es)

          inflammatory response
          G protein-coupled receptor signaling pathway
          male sex determination
          sleep
          adenosine metabolic process
          regulation of cytosolic calcium ion concentration
          positive regulation of penile erection
          cellular response to prostaglandin D stimulus
          prostaglandin J receptor activity
          prostaglandin receptor activity
          prostaglandin D receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245331 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43549 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49205 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10779521 - -
          Affymetrix Rat Gene 1.0 ST Array 10782800 - -
          Affymetrix Rat Gene 1.0 ST Array 10779522 - -
          Affymetrix Rat Gene 1.1 ST Array 10779520 - -
          Affymetrix Rat Gene 1.1 ST Array 10782799 - -
          Rat Genome 230 2.0 Array 1387523_PM_at - -
          Rat Genome 230 2.0 Array 1387523_at - -
          Affymetrix Rat Clariom S Assay PSR1500008919.rn.1 TC1500001403.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500008920.rn.1 TC1500001403.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500008918.rn.1 TC1500001403.rn.1 RTA TC
          Rat Expression Set 230 1387523_PM_at - -
          Rat Expression Set 230 1387523_at - -
          Affymetrix Rat Gene 1.0 ST Array 10782799 - -
          Affymetrix Rat Gene 1.0 ST Array 10779520 - -
          Rat Genome U34 Set U92289_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351502 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0