Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01217703_g1
          See other TFDP1 GE Assays ›
          Gene Symbol:
          Tfdp1
          Gene Name:
          transcription factor Dp-1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 81088640 - 81127244 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01217703_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tfdp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025718.1 NP_001020889.1 8 - 9 1063 57
          XM_006253451.2 XP_006253513.1 9 - 10 1328 57
          XM_008771397.1 XP_008769619.1 10 - 11 1331 57
          XM_008771398.1 XP_008769620.1 10 - 11 1410 57
          XM_008771399.1 XP_008769621.1 10 - 11 1238 57
          XM_008771400.1 XP_008769622.1 9 - 10 1364 57
          GenBank mRNA BC098789.1 - 8 - 9 1063 57

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tfdp1

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cell cycle
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          epidermis development
          anoikis
          positive regulation of transcription, DNA-templated
          regulation of DNA biosynthetic process
          DNA binding
          DNA-binding transcription factor activity
          transcription coactivator activity
          transcription factor binding
          protein domain specific binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P247139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106938 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83503 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66936 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44538 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87861 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111940 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P247139 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P247139 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P247139 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P249606 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P247139 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44278 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40512 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73538 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95627 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32326 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10792755 - -
          Affymetrix Rat Gene 1.0 ST Array 10792753 - -
          Affymetrix Rat Gene 1.0 ST Array 10792757 - -
          Affymetrix Rat Gene 1.0 ST Array 10792752 - -
          Affymetrix Rat Gene 1.0 ST Array 10792761 - -
          Affymetrix Rat Gene 1.0 ST Array 10792756 - -
          Affymetrix Rat Gene 1.0 ST Array 10792754 - -
          Affymetrix Rat Gene 1.0 ST Array 10792759 - -
          Affymetrix Rat Gene 1.0 ST Array 10792758 - -
          Affymetrix Rat Gene 1.1 ST Array 10792751 - -
          Rat Genome 230 2.0 Array 1388867_PM_at - -
          Rat Genome 230 2.0 Array 1388867_at - -
          Affymetrix HT Rat Focus Array 1388867_PM_at - -
          Affymetrix HT Rat Focus Array 1388867_at - -
          Affymetrix Rat Clariom S Assay PSR1600007255.rn.1 TC1600000964.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600007254.rn.1 TC1600000964.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600007253.rn.1 TC1600000964.rn.1 RTA TC
          Rat Expression Set 230 1388867_at - -
          Rat Expression Set 230 1388867_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10792751 - -
          Rat Genome U34 Set rc_AI227852_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21084743 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360118 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0