Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01243220_g1
          See other DDX19A GE Assays ›
          Gene Symbol:
          Ddx19a Ddx19b
          Gene Name:
          DEAD (Asp-Glu-Ala-Asp) box polypeptide 19a
          DEAD (Asp-Glu-Ala-As) box polypeptide 19B
          Gene Aliases:
          Ddx19, ZD10B
          Zd10a
          Chromosome Location:
          Chr.19: 43251510 - 43271866 on Build Rnor_6.0
          Chromosome Location:
          Chr.19: 43217199 - 43246196 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01243220_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ddx19a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001005381.2 NP_001005381.2 8 - 9 854 65
          XM_008772490.1 XP_008770712.1 640 65
          GenBank mRNA AY303540.1 - 872 65
          BC079094.1 - 8 - 9 825 65
          Gene Symbol: Ddx19b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001005895.2 NP_001005895.2 8 - 9 930 65
          XM_008772534.1 XP_008770756.1 8 - 9 844 65
          GenBank mRNA AY751756.1 - 8 - 9 872 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000604.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ddx19a
          Click here to find siRNAs for Gene Ddx19b

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          response to zinc ion
          positive regulation of apoptotic process
          nucleic acid binding
          helicase activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P480636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270608 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116274 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158843 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12112 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33427 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164351 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P048581 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P480636 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P480636 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P048581 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P048581 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P480636 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P480636 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50654 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42859 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64282 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90975 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63219 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10811057 - -
          Affymetrix Rat Gene 1.0 ST Array 10811055 - -
          Affymetrix Rat Gene 1.0 ST Array 10811054 - -
          Affymetrix Rat Gene 1.0 ST Array 10811042 - -
          Affymetrix Rat Gene 1.0 ST Array 10811058 - -
          Affymetrix Rat Gene 1.0 ST Array 10811052 - -
          Affymetrix Rat Gene 1.0 ST Array 10811050 - -
          Affymetrix Rat Gene 1.0 ST Array 10811053 - -
          Affymetrix Rat Gene 1.0 ST Array 10811049 - -
          Affymetrix Rat Gene 1.0 ST Array 10811051 - -
          Affymetrix Rat Gene 1.0 ST Array 10811046 - -
          Affymetrix Rat Gene 1.0 ST Array 10811055 - -
          Affymetrix Rat Gene 1.0 ST Array 10811048 - -
          Affymetrix Rat Gene 1.0 ST Array 10811057 - -
          Affymetrix Rat Gene 1.0 ST Array 10811044 - -
          Affymetrix Rat Gene 1.0 ST Array 10811047 - -
          Affymetrix Rat Gene 1.0 ST Array 10811056 - -
          Affymetrix Rat Gene 1.0 ST Array 10811052 - -
          Affymetrix Rat Gene 1.0 ST Array 10811056 - -
          Affymetrix Rat Gene 1.0 ST Array 10811043 - -
          Affymetrix Rat Gene 1.0 ST Array 10811045 - -
          Affymetrix Rat Gene 1.0 ST Array 10811058 - -
          Affymetrix Rat Gene 1.1 ST Array 10811041 - -
          Rat Genome 230 2.0 Array 1383559_PM_at - -
          Rat Genome 230 2.0 Array 1396661_PM_at - -
          Rat Genome 230 2.0 Array 1396661_at - -
          Rat Genome 230 2.0 Array 1378934_at - -
          Rat Genome 230 2.0 Array 1378934_PM_at - -
          Rat Genome 230 2.0 Array 1388814_PM_at - -
          Rat Genome 230 2.0 Array 1388814_at - -
          Rat Genome 230 2.0 Array 1383559_at - -
          Rat Genome 230 2.0 Array 1381134_at - -
          Rat Genome 230 2.0 Array 1381134_PM_at - -
          Affymetrix HT Rat Focus Array 1380012_at - -
          Affymetrix HT Rat Focus Array 1381616_PM_at - -
          Affymetrix HT Rat Focus Array 1396661_PM_at - -
          Affymetrix HT Rat Focus Array 1381616_at - -
          Affymetrix HT Rat Focus Array 1388814_at - -
          Affymetrix HT Rat Focus Array 1388814_PM_at - -
          Affymetrix HT Rat Focus Array 1388843_PM_at - -
          Affymetrix HT Rat Focus Array 1396661_at - -
          Affymetrix HT Rat Focus Array 1380012_PM_at - -
          Affymetrix HT Rat Focus Array 1388843_at - -
          Affymetrix Rat Clariom S Assay PSR1900005148.rn.1 TC1900000604.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900005168.rn.1 TC1900000604.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900005169.rn.1 TC1900000604.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900005173.rn.1 TC1900000604.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900005174.rn.1 TC1900000604.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900005150.rn.1 TC1900000604.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900005167.rn.1 TC1900000604.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900005172.rn.1 TC1900000604.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900005146.rn.1 TC1900000604.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900005147.rn.1 TC1900000604.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900005151.rn.1 TC1900000604.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900005145.rn.1 TC1900000604.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900005149.rn.1 TC1900000604.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900005171.rn.1 TC1900000604.rn.1 RTA TC
          Rat Expression Set 230 1388814_at - -
          Rat Expression Set 230 1388814_PM_at - -
          Rat Expression Set 230 1396661_at - -
          Rat Expression Set 230 1383559_PM_at - -
          Rat Expression Set 230 1396661_PM_at - -
          Rat Expression Set 230 1383559_at - -
          Affymetrix Rat Gene 1.0 ST Array 10811041 - -
          Rat Genome U34 Set rc_AI045477_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21304845 - -
          ABI Rat Genome Survey Microarray 22182717 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354324 - -
          RatRef-12 Expression BeadChip ILMN_69649 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0