Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01269329_m1
          See other CTDSPL2 GE Assays ›
          Gene Symbol:
          Ctdspl2
          Gene Name:
          CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase like 2
          Gene Aliases:
          RGD1309219
          Chromosome Location:
          Chr.3: 113906803 - 113957289 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01269329_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ctdspl2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014048.1 NP_001014070.1 12 - 13 1433 153
          GenBank mRNA BC083672.1 - 12 - 13 1433 153

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300001579.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ctdspl2

          Gene Ontology Categories:

          Function(s) Process(es)

          protein dephosphorylation
          dephosphorylation
          negative regulation of BMP signaling pathway
          positive regulation of protein export from nucleus
          phosphoprotein phosphatase activity
          phosphatase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P047152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904630 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172021 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47215 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162715 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7972 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43108 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135190 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160048 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54320 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118899 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166943 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39605 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047152 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047152 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P047152 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97677 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15834 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86223 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67950 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25663 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10839217 - -
          Affymetrix Rat Gene 1.0 ST Array 10839221 - -
          Affymetrix Rat Gene 1.0 ST Array 10839218 - -
          Affymetrix Rat Gene 1.0 ST Array 10839216 - -
          Affymetrix Rat Gene 1.0 ST Array 10839214 - -
          Affymetrix Rat Gene 1.0 ST Array 10839210 - -
          Affymetrix Rat Gene 1.0 ST Array 10839212 - -
          Affymetrix Rat Gene 1.0 ST Array 10839219 - -
          Affymetrix Rat Gene 1.0 ST Array 10839215 - -
          Affymetrix Rat Gene 1.0 ST Array 10839211 - -
          Affymetrix Rat Gene 1.0 ST Array 10839209 - -
          Affymetrix Rat Gene 1.0 ST Array 10839213 - -
          Affymetrix Rat Gene 1.0 ST Array 10839220 - -
          Affymetrix Rat Gene 1.1 ST Array 10839208 - -
          Rat Genome 230 2.0 Array 1383179_at - -
          Rat Genome 230 2.0 Array 1383179_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300010987.rn.1 TC0300001579.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10839208 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371704 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0