Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01310378_g1
          See other NKIRAS1 GE Assays ›
          Gene Symbol:
          Nkiras1
          Gene Name:
          NFKB inhibitor interacting Ras-like 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 8179090 - 8188720 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01310378_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nkiras1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107252.2 NP_001100722.1 4 - 5 642 95
          XM_006251698.2 XP_006251760.1 3 - 4 616 95
          XM_008770514.1 XP_008768736.1 4 - 5 770 95
          GenBank mRNA BC169080.1 - 4 - 5 642 95
          FQ211880.1 - 527 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001321.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          small GTPase mediated signal transduction
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P259125 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P710324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54798 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178869 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P710324 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P259125 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P259125 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P710324 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P259125 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P710324 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P259125 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P710324 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89085 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35297 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92571 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10782452 - -
          Affymetrix Rat Gene 1.0 ST Array 10782451 - -
          Affymetrix Rat Gene 1.0 ST Array 10782453 - -
          Affymetrix Rat Gene 1.1 ST Array 10782450 - -
          Rat Genome 230 2.0 Array 1394590_at - -
          Rat Genome 230 2.0 Array 1396092_PM_at - -
          Rat Genome 230 2.0 Array 1396092_at - -
          Rat Genome 230 2.0 Array 1393847_at - -
          Rat Genome 230 2.0 Array 1393847_PM_at - -
          Rat Genome 230 2.0 Array 1394590_PM_at - -
          Affymetrix HT Rat Focus Array 1393847_PM_at - -
          Affymetrix HT Rat Focus Array 1394590_PM_at - -
          Affymetrix HT Rat Focus Array 1393847_at - -
          Affymetrix HT Rat Focus Array 1396092_at - -
          Affymetrix HT Rat Focus Array 1394590_at - -
          Affymetrix HT Rat Focus Array 1396092_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1500008338.rn.1 TC1500001321.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500008337.rn.1 TC1500001321.rn.1 RTA TC
          Rat Expression Set 230 1394590_PM_at - -
          Rat Expression Set 230 1396092_PM_at - -
          Rat Expression Set 230 1396092_at - -
          Rat Expression Set 230 1393847_at - -
          Rat Expression Set 230 1393847_PM_at - -
          Rat Expression Set 230 1394590_at - -
          Affymetrix Rat Gene 1.0 ST Array 10782450 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21405822 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361699 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline