Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01400171_m1
          See other DAW1 GE Assays ›
          Gene Symbol:
          Daw1
          Gene Name:
          dynein assembly factor with WDR repeat domains 1
          Gene Aliases:
          Wdr69
          Chromosome Location:
          Chr.9: 88964530 - 89001568 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01400171_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Daw1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025025.1 NP_001020196.1 11 - 12 1092 64
          GenBank mRNA BC079402.1 - 11 - 12 1075 64
          BC091226.1 - 11 - 12 1092 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900001002.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Daw1

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P424614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885882 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150708 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147254 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143932 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14560 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P424614 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P424614 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P424614 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P424614 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1701 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3377 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66097 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10924793 - -
          Affymetrix Rat Gene 1.0 ST Array 10924795 - -
          Affymetrix Rat Gene 1.0 ST Array 10924787 - -
          Affymetrix Rat Gene 1.0 ST Array 10924792 - -
          Affymetrix Rat Gene 1.0 ST Array 10924788 - -
          Affymetrix Rat Gene 1.0 ST Array 10924791 - -
          Affymetrix Rat Gene 1.0 ST Array 10924786 - -
          Affymetrix Rat Gene 1.0 ST Array 10924796 - -
          Affymetrix Rat Gene 1.0 ST Array 10924789 - -
          Affymetrix Rat Gene 1.0 ST Array 10924790 - -
          Affymetrix Rat Gene 1.0 ST Array 10924794 - -
          Affymetrix Rat Gene 1.0 ST Array 10924797 - -
          Affymetrix Rat Gene 1.1 ST Array 10924785 - -
          Affymetrix Rat Clariom S Assay PSR0900008482.rn.1 TC0900001002.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10924785 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21534190 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372870 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline