Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01400276_m1
          See other PDE6D GE Assays ›
          Gene Symbol:
          Pde6d
          Gene Name:
          phosphodiesterase 6D, cGMP-specific, rod, delta
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 93562098 - 93607076 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01400276_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pde6d
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108806.1 NP_001102276.1 4 - 5 504 93
          XM_006245413.2 XP_006245475.1 4 - 5 552 93
          GenBank mRNA FQ216262.1 - 2 - 3 126 93
          FQ227000.1 - 2 - 3 126 93
          FQ234983.1 - 2 - 3 126 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900002399.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus Rat Phosphodiesterase 1700 Inventoried Rat Phosphodiesterase Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          visual perception
          metabolic process
          negative regulation of catalytic activity
          3',5'-cyclic-nucleotide phosphodiesterase activity
          GTPase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P405821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85546 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137854 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58976 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80931 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135534 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131717 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34097 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P405821 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P405821 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P405821 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P405821 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7115 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10128 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33200 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10929601 - -
          Affymetrix Rat Gene 1.0 ST Array 10929603 - -
          Affymetrix Rat Gene 1.0 ST Array 10929602 - -
          Affymetrix Rat Gene 1.0 ST Array 10929605 - -
          Affymetrix Rat Gene 1.0 ST Array 10929604 - -
          Affymetrix Rat Gene 1.1 ST Array 10929600 - -
          Rat Genome 230 2.0 Array 1388512_at - -
          Rat Genome 230 2.0 Array 1388512_PM_at - -
          Affymetrix HT Rat Focus Array 1388512_PM_at - -
          Affymetrix HT Rat Focus Array 1388512_at - -
          Affymetrix Rat Clariom S Assay PSR0900019650.rn.1 TC0900002399.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900019649.rn.1 TC0900002399.rn.1 RTA TC
          Rat Expression Set 230 1388512_at - -
          Rat Expression Set 230 1388512_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10929600 - -
          Rat Genome U34 Set rc_AA875045_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21140728 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360087 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0