Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01402153_m1
          See other ARHGEF6 GE Assays ›
          Gene Symbol:
          Arhgef6
          Gene Name:
          Rac/Cdc42 guanine nucleotide exchange factor (GEF) 6
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 159722031 - 159841335 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01402153_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arhgef6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001005565.1 NP_001005565.1 21 - 22 2177 165
          XM_008773651.1 XP_008771873.1 20 - 21 2687 165
          XM_008773652.1 XP_008771874.1 20 - 21 2582 165
          XM_008773653.1 XP_008771875.1 19 - 20 2560 165
          XM_008773654.1 XP_008771876.1 20 - 21 1999 165
          GenBank mRNA AY725848.1 - 21 - 22 2177 165

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00002674.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arhgef6

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          lamellipodium assembly
          intracellular signal transduction
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P072402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48734 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142591 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130934 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18173 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64911 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118625 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33147 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15420 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104564 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126255 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144077 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142191 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102327 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22346 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131725 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P072402 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P072402 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P072402 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P539175 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P232727 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94902 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9571 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60939 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75373 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18652 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82625 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17424 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11191 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10939875 - -
          Affymetrix Rat Gene 1.0 ST Array 10939881 - -
          Affymetrix Rat Gene 1.0 ST Array 10939877 - -
          Affymetrix Rat Gene 1.0 ST Array 10939887 - -
          Affymetrix Rat Gene 1.0 ST Array 10939883 - -
          Affymetrix Rat Gene 1.0 ST Array 10939873 - -
          Affymetrix Rat Gene 1.0 ST Array 10939885 - -
          Affymetrix Rat Gene 1.0 ST Array 10939868 - -
          Affymetrix Rat Gene 1.0 ST Array 10939879 - -
          Affymetrix Rat Gene 1.0 ST Array 10939874 - -
          Affymetrix Rat Gene 1.0 ST Array 10939876 - -
          Affymetrix Rat Gene 1.0 ST Array 10939878 - -
          Affymetrix Rat Gene 1.0 ST Array 10939870 - -
          Affymetrix Rat Gene 1.0 ST Array 10939884 - -
          Affymetrix Rat Gene 1.0 ST Array 10939886 - -
          Affymetrix Rat Gene 1.0 ST Array 10939872 - -
          Affymetrix Rat Gene 1.0 ST Array 10939880 - -
          Affymetrix Rat Gene 1.0 ST Array 10939871 - -
          Affymetrix Rat Gene 1.0 ST Array 10939882 - -
          Affymetrix Rat Gene 1.0 ST Array 10939869 - -
          Affymetrix Rat Gene 1.1 ST Array 10939866 - -
          Affymetrix Rat Clariom S Assay PSR0X00016871.rn.1 TC0X00002674.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00016869.rn.1 TC0X00002674.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10939866 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21716863 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376939 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0