Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01402438_m1
          See other OXCT1 GE Assays ›
          Gene Symbol:
          Oxct1
          Gene Name:
          3-oxoacid CoA transferase 1
          Gene Aliases:
          SCOT
          Chromosome Location:
          Chr.2: 53859738 - 54007733 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01402438_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Oxct1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001127580.1 NP_001121052.1 14 - 15 1440 79
          GenBank mRNA AY724483.1 - 14 - 15 1394 79
          BC166478.1 - 14 - 15 1440 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200000484.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          brain development
          heart development
          response to nutrient
          response to xenobiotic stimulus
          response to hormone
          response to activity
          positive regulation of insulin secretion involved in cellular response to glucose stimulus
          ketone catabolic process
          response to starvation
          response to ethanol
          cellular ketone body metabolic process
          ketone body catabolic process
          adipose tissue development
          3-oxoacid CoA-transferase activity
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P746868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556686 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158089 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136414 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114639 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98701 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134511 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27178 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163916 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169992 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82666 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128538 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155999 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3819 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123082 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P746868 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P746868 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P746868 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P503809 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10813285 - -
          Affymetrix Rat Gene 1.0 ST Array 10813278 - -
          Affymetrix Rat Gene 1.0 ST Array 10813284 - -
          Affymetrix Rat Gene 1.0 ST Array 10813276 - -
          Affymetrix Rat Gene 1.0 ST Array 10813280 - -
          Affymetrix Rat Gene 1.0 ST Array 10813281 - -
          Affymetrix Rat Gene 1.0 ST Array 10813283 - -
          Affymetrix Rat Gene 1.0 ST Array 10813277 - -
          Affymetrix Rat Gene 1.0 ST Array 10813279 - -
          Affymetrix Rat Gene 1.0 ST Array 10813275 - -
          Affymetrix Rat Gene 1.0 ST Array 10813274 - -
          Affymetrix Rat Gene 1.0 ST Array 10813273 - -
          Affymetrix Rat Gene 1.0 ST Array 10813272 - -
          Affymetrix Rat Gene 1.0 ST Array 10813282 - -
          Affymetrix Rat Gene 1.1 ST Array 10813271 - -
          Rat Genome 230 2.0 Array 1371421_at - -
          Rat Genome 230 2.0 Array 1371421_PM_at - -
          Affymetrix HT Rat Focus Array 1371421_PM_at - -
          Affymetrix HT Rat Focus Array 1371421_at - -
          Affymetrix Rat Clariom S Assay PSR0200002807.rn.1 TC0200000484.rn.1 RTA TC
          Rat Expression Set 230 1371421_PM_at - -
          Rat Expression Set 230 1371421_at - -
          Affymetrix Rat Gene 1.0 ST Array 10813271 - -
          Rat Genome U34 Set rc_AI105080_at - -
          Rat Genome U34 Set rc_AA819087_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21257719 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0