Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01402688_m1
          See other CAPSL GE Assays ›
          Gene Symbol:
          Capsl
          Gene Name:
          calcyphosine-like
          Gene Aliases:
          RGD1308776
          Chromosome Location:
          Chr.2: 58724855 - 58756261 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01402688_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Capsl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106417.1 NP_001099887.1 4 - 5 666 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200000547.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Process(es)

          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P492173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656901 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166996 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40478 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62412 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P492173 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P492173 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P492173 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P492173 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57945 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10813626 - -
          Affymetrix Rat Gene 1.0 ST Array 10813625 - -
          Affymetrix Rat Gene 1.0 ST Array 10813627 - -
          Affymetrix Rat Gene 1.0 ST Array 10813624 - -
          Affymetrix Rat Gene 1.0 ST Array 10813623 - -
          Affymetrix Rat Gene 1.1 ST Array 10813622 - -
          Affymetrix Rat Clariom S Assay PSR0200003427.rn.1 TC0200000547.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10813622 - -
          Rat Genome U34 Set rc_AI031051_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21336221 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369079 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0