Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01404761_m1
          See other ABCF3 GE Assays ›
          Gene Symbol:
          Abcf3
          Gene Name:
          ATP-binding cassette, subfamily F (GCN20), member 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 84026206 - 84037938 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01404761_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Abcf3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011896.1 NP_001011896.1 14 - 15 1471 77
          GenBank mRNA BC082042.1 - 14 - 15 1471 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100001938.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Abcf3

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          defense response to virus
          ATP binding
          ATP hydrolysis activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P506751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351702 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138712 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P506751 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P506751 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P506751 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P506751 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53947 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5499 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10755441 - -
          Affymetrix Rat Gene 1.0 ST Array 10755427 - -
          Affymetrix Rat Gene 1.0 ST Array 10755434 - -
          Affymetrix Rat Gene 1.0 ST Array 10755437 - -
          Affymetrix Rat Gene 1.0 ST Array 10755445 - -
          Affymetrix Rat Gene 1.0 ST Array 10755428 - -
          Affymetrix Rat Gene 1.0 ST Array 10755443 - -
          Affymetrix Rat Gene 1.0 ST Array 10755435 - -
          Affymetrix Rat Gene 1.0 ST Array 10755429 - -
          Affymetrix Rat Gene 1.0 ST Array 10755444 - -
          Affymetrix Rat Gene 1.0 ST Array 10755431 - -
          Affymetrix Rat Gene 1.0 ST Array 10755438 - -
          Affymetrix Rat Gene 1.0 ST Array 10755432 - -
          Affymetrix Rat Gene 1.0 ST Array 10755442 - -
          Affymetrix Rat Gene 1.0 ST Array 10755436 - -
          Affymetrix Rat Gene 1.0 ST Array 10755440 - -
          Affymetrix Rat Gene 1.0 ST Array 10755430 - -
          Affymetrix Rat Gene 1.0 ST Array 10755446 - -
          Affymetrix Rat Gene 1.0 ST Array 10755433 - -
          Affymetrix Rat Gene 1.0 ST Array 10755439 - -
          Affymetrix Rat Gene 1.0 ST Array 10755447 - -
          Affymetrix Rat Gene 1.1 ST Array 10755426 - -
          Rat Genome 230 2.0 Array 1371456_PM_at - -
          Rat Genome 230 2.0 Array 1371456_at - -
          Rat Genome 230 2.0 Array 1377280_at - -
          Rat Genome 230 2.0 Array 1377280_PM_at - -
          Affymetrix HT Rat Focus Array 1371456_PM_at - -
          Affymetrix HT Rat Focus Array 1377280_PM_at - -
          Affymetrix HT Rat Focus Array 1377280_at - -
          Affymetrix HT Rat Focus Array 1371456_at - -
          Affymetrix Rat Clariom S Assay PSR1100012643.rn.1 TC1100001938.rn.1 RTA TC
          Rat Expression Set 230 1371456_PM_at - -
          Rat Expression Set 230 1377280_PM_at - -
          Rat Expression Set 230 1371456_at - -
          Rat Expression Set 230 1377280_at - -
          Affymetrix Rat Gene 1.0 ST Array 10755426 - -
          Rat Genome U34 Set rc_AI169668_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376754 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0