Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01405422_m1
          See other LOC100911837 GE Assays ›
          Gene Symbol:
          LOC100911837 Hira
          Gene Name:
          protein HIRA-like
          histone cell cycle regulator
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.11: 85802536 - 85896668 on Build Rnor_6.0
          Chromosome Location:
          Chr.11: 86168196 - 86276430 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01405422_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100911837
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008768905.1 XP_008767127.1 3599 72
          XM_008768906.1 XP_008767128.1 2143 72
          XM_008768907.1 XP_008767129.1 2072 72
          XM_008768908.1 XP_008767130.1 2670 72
          XM_008768909.1 XP_008767131.1 1614 72
          XM_008768910.1 XP_008767132.1 1692 72
          Gene Symbol: Hira
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001135760.1 NP_001129232.1 14 - 15 2312 72
          XM_006248639.2 XP_006248701.1 15 - 16 1613 72
          XM_006248641.2 XP_006248703.1 12 - 13 1692 72
          XM_008768893.1 XP_008767115.1 14 - 15 4115 72
          XM_008768894.1 XP_008767116.1 14 - 15 2305 72
          XM_008768895.1 XP_008767117.1 14 - 15 2422 72
          XM_008768896.1 XP_008767118.1 13 - 14 2072 72
          XM_008768897.1 XP_008767119.1 15 - 16 2012 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100001967.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin organization
          regulation of transcription, DNA-templated
          osteoblast differentiation
          DNA replication-independent nucleosome assembly
          gastrulation
          biological_process
          muscle cell differentiation
          chromatin binding
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P020079 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P020078 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P020078 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P020078 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P138363 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10756016 - -
          Affymetrix Rat Gene 1.0 ST Array 10756033 - -
          Affymetrix Rat Gene 1.0 ST Array 10756036 - -
          Affymetrix Rat Gene 1.0 ST Array 10756030 - -
          Affymetrix Rat Gene 1.0 ST Array 10756017 - -
          Affymetrix Rat Gene 1.0 ST Array 10756019 - -
          Affymetrix Rat Gene 1.0 ST Array 10756023 - -
          Affymetrix Rat Gene 1.0 ST Array 10756032 - -
          Affymetrix Rat Gene 1.0 ST Array 10756027 - -
          Affymetrix Rat Gene 1.0 ST Array 10756015 - -
          Affymetrix Rat Gene 1.0 ST Array 10756029 - -
          Affymetrix Rat Gene 1.0 ST Array 10756013 - -
          Affymetrix Rat Gene 1.0 ST Array 10756020 - -
          Affymetrix Rat Gene 1.0 ST Array 10756028 - -
          Affymetrix Rat Gene 1.0 ST Array 10756035 - -
          Affymetrix Rat Gene 1.0 ST Array 10756034 - -
          Affymetrix Rat Gene 1.0 ST Array 10756025 - -
          Affymetrix Rat Gene 1.0 ST Array 10756021 - -
          Affymetrix Rat Gene 1.0 ST Array 10756026 - -
          Affymetrix Rat Gene 1.0 ST Array 10756031 - -
          Affymetrix Rat Gene 1.0 ST Array 10756022 - -
          Affymetrix Rat Gene 1.0 ST Array 10756014 - -
          Affymetrix Rat Gene 1.0 ST Array 10756018 - -
          Affymetrix Rat Gene 1.0 ST Array 10756012 - -
          Affymetrix Rat Gene 1.1 ST Array 10756011 - -
          Rat Genome 230 2.0 Array 1382316_at - -
          Rat Genome 230 2.0 Array 1382316_PM_at - -
          Rat Genome 230 2.0 Array 1385811_at - -
          Rat Genome 230 2.0 Array 1385811_PM_at - -
          Affymetrix HT Rat Focus Array 1385811_at - -
          Affymetrix HT Rat Focus Array 1382316_PM_at - -
          Affymetrix HT Rat Focus Array 1382316_at - -
          Affymetrix HT Rat Focus Array 1385811_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1100012893.rn.1 TC1100001967.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100012894.rn.1 TC1100001967.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100012896.rn.1 TC1100001967.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100012899.rn.1 TC1100001967.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100012897.rn.1 TC1100001967.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100012892.rn.1 TC1100001967.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100012898.rn.1 TC1100001967.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100012890.rn.1 TC1100001967.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100012900.rn.1 TC1100001967.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100012891.rn.1 TC1100001967.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100012895.rn.1 TC1100001967.rn.1 RTA TC
          Rat Expression Set 230 1385811_PM_at - -
          Rat Expression Set 230 1382316_PM_at - -
          Rat Expression Set 230 1385811_at - -
          Rat Expression Set 230 1382316_at - -
          Affymetrix Rat Gene 1.0 ST Array 10756011 - -
          Rat Genome U34 Set rc_AI177968_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22422992 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372178 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0