Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01405902_m1
          See other ZFP35 GE Assays ›
          Gene Symbol:
          Zfp35
          Gene Name:
          zinc finger protein 35
          Gene Aliases:
          Zfp239
          Chromosome Location:
          Chr.18: 15883182 - 15898803 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01405902_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp35
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013141.1 NP_001013159.1 2 - 3 361 85
          GenBank mRNA BC089850.1 - 2 - 3 361 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800000148.rn.1

          Additional Information:

          This assay is in the 5'UTR region of the BC089850.1, NM_001013141.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp35

          Gene Ontology Categories:

          Process(es)

          nucleic acid binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P140956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72681 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35621 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P140956 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P140956 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P140956 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103692 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14988 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97716 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10800495 - -
          Affymetrix Rat Gene 1.0 ST Array 10800494 - -
          Affymetrix Rat Gene 1.0 ST Array 10800496 - -
          Affymetrix Rat Gene 1.1 ST Array 10800493 - -
          Affymetrix Rat Clariom S Assay PSR1800001003.rn.1 TC1800000148.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800001001.rn.1 TC1800000148.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800001002.rn.1 TC1800000148.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800001006.rn.1 TC1800000148.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800001004.rn.1 TC1800000148.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800001005.rn.1 TC1800000148.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10800493 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21545729 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650495 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0