Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01406673_m1
          See other PKD2L2 GE Assays ›
          Gene Symbol:
          Pkd2l2
          Gene Name:
          polycystic kidney disease 2-like 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 27190661 - 27225488 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01406673_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pkd2l2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106156.1 NP_001099626.1 13 - 14 1821 67
          XM_008772026.1 XP_008770248.1 13 - 14 2134 67
          XM_008772027.1 XP_008770249.1 13 - 14 2068 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800000259.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          detection of mechanical stimulus
          calcium ion transmembrane transport
          calcium channel activity
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P173289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417735 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175028 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75012 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57179 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44536 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49209 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P173289 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P173289 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P173289 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P173289 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10800871 - -
          Affymetrix Rat Gene 1.0 ST Array 10800877 - -
          Affymetrix Rat Gene 1.0 ST Array 10800868 - -
          Affymetrix Rat Gene 1.0 ST Array 10800880 - -
          Affymetrix Rat Gene 1.0 ST Array 10800869 - -
          Affymetrix Rat Gene 1.0 ST Array 10800873 - -
          Affymetrix Rat Gene 1.0 ST Array 10800874 - -
          Affymetrix Rat Gene 1.0 ST Array 10800872 - -
          Affymetrix Rat Gene 1.0 ST Array 10800870 - -
          Affymetrix Rat Gene 1.0 ST Array 10800879 - -
          Affymetrix Rat Gene 1.0 ST Array 10800878 - -
          Affymetrix Rat Gene 1.0 ST Array 10800875 - -
          Affymetrix Rat Gene 1.0 ST Array 10800876 - -
          Affymetrix Rat Gene 1.0 ST Array 10800867 - -
          Affymetrix Rat Gene 1.0 ST Array 10800881 - -
          Affymetrix Rat Gene 1.1 ST Array 10800865 - -
          Rat Genome 230 2.0 Array 1386750_PM_at - -
          Rat Genome 230 2.0 Array 1386750_at - -
          Affymetrix HT Rat Focus Array 1386750_PM_at - -
          Affymetrix HT Rat Focus Array 1386750_at - -
          Affymetrix Rat Clariom S Assay PSR1800001901.rn.1 TC1800000259.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800001900.rn.1 TC1800000259.rn.1 RTA TC
          Rat Expression Set 230 1386750_PM_at - -
          Rat Expression Set 230 1386750_at - -
          Affymetrix Rat Gene 1.0 ST Array 10800865 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22179744 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0