Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01407196_m1
          See other MORC3 GE Assays ›
          Gene Symbol:
          Morc3
          Gene Name:
          MORC family CW-type zinc finger 3
          Gene Aliases:
          Zcwcc3
          Chromosome Location:
          Chr.11: 34051927 - 34094697 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01407196_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Morc3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107109.1 NP_001100579.1 15 - 16 2548 64
          XM_006248060.1 XP_006248122.1 16 - 17 2881 64
          XM_006248061.2 XP_006248123.1 15 - 16 2807 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100000315.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          cell aging
          post-embryonic development
          peptidyl-serine phosphorylation
          negative regulation of fibroblast proliferation
          protein stabilization
          maintenance of protein location in nucleus
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P322001 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P133696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396724 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36139 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101577 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71436 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168140 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56013 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P133696 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P322001 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P133696 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P322001 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P133696 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P322001 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P521609 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P322001 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19233 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63173 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25377 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10750377 - -
          Affymetrix Rat Gene 1.0 ST Array 10750387 - -
          Affymetrix Rat Gene 1.0 ST Array 10750382 - -
          Affymetrix Rat Gene 1.0 ST Array 10750381 - -
          Affymetrix Rat Gene 1.0 ST Array 10750378 - -
          Affymetrix Rat Gene 1.0 ST Array 10750384 - -
          Affymetrix Rat Gene 1.0 ST Array 10750385 - -
          Affymetrix Rat Gene 1.0 ST Array 10750380 - -
          Affymetrix Rat Gene 1.0 ST Array 10750390 - -
          Affymetrix Rat Gene 1.0 ST Array 10750389 - -
          Affymetrix Rat Gene 1.0 ST Array 10750386 - -
          Affymetrix Rat Gene 1.0 ST Array 10750388 - -
          Affymetrix Rat Gene 1.0 ST Array 10750375 - -
          Affymetrix Rat Gene 1.0 ST Array 10750376 - -
          Affymetrix Rat Gene 1.0 ST Array 10750383 - -
          Affymetrix Rat Gene 1.0 ST Array 10750374 - -
          Affymetrix Rat Gene 1.1 ST Array 10750373 - -
          Rat Genome 230 2.0 Array 1390142_at - -
          Rat Genome 230 2.0 Array 1390142_PM_at - -
          Affymetrix HT Rat Focus Array 1390142_PM_at - -
          Affymetrix HT Rat Focus Array 1390142_at - -
          Affymetrix Rat Clariom S Assay PSR1100001431.rn.1 TC1100000315.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100001432.rn.1 TC1100000315.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100001429.rn.1 TC1100000315.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100001430.rn.1 TC1100000315.rn.1 RTA TC
          Rat Expression Set 230 1390142_at - -
          Rat Expression Set 230 1390142_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10750373 - -
          Rat Genome U34 Set rc_AI177734_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22111809 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360936 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0