Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01407921_g1
          See other AMD1 GE Assays ›
          Gene Symbol:
          Amd1
          Gene Name:
          adenosylmethionine decarboxylase 1
          Gene Aliases:
          Amd1a, Amd1b
          Chromosome Location:
          Chr.20: 45384000 - 45399694 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01407921_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Amd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031011.3 NP_112273.3 7 - 8 1182 72
          GenBank mRNA AY724509.1 - 7 - 8 865 72
          BC061532.1 - 7 - 8 1185 72
          FQ232080.1 - 7 - 8 1002 72
          FQ234346.1 - 7 - 8 1005 72
          FQ234398.1 - 2 - 3 1351 72
          M34464.1 - 7 - 8 1128 72

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Amd1

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          S-adenosylmethioninamine biosynthetic process
          polyamine metabolic process
          spermine biosynthetic process
          spermidine biosynthetic process
          S-adenosylmethionine metabolic process
          adenosylmethionine decarboxylase activity
          putrescine binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P595591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688001 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36267 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137754 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P595591 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P595591 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P595591 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P595591 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1020324 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102195 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4170 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10833662 - -
          Affymetrix Rat Gene 1.0 ST Array 10833666 - -
          Affymetrix Rat Gene 1.0 ST Array 10833661 - -
          Affymetrix Rat Gene 1.0 ST Array 10833665 - -
          Affymetrix Rat Gene 1.0 ST Array 10833664 - -
          Affymetrix Rat Gene 1.0 ST Array 10833663 - -
          Affymetrix Rat Gene 1.0 ST Array 10833667 - -
          Affymetrix Rat Gene 1.0 ST Array 10833660 - -
          Affymetrix Rat Gene 1.1 ST Array 10833659 - -
          Rat Genome 230 2.0 Array 1367933_PM_at - -
          Rat Genome 230 2.0 Array 1367933_at - -
          Affymetrix HT Rat Focus Array 1367933_at - -
          Affymetrix HT Rat Focus Array 1367933_PM_at - -
          Affymetrix Rat Clariom S Assay PSR2000013723.rn.1 TC2000001692.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000013724.rn.1 TC2000001692.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000013725.rn.1 TC2000001692.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000013727.rn.1 TC2000001692.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000013722.rn.1 TC2000001692.rn.1 RTA TC
          Rat Expression Set 230 1367933_at - -
          Rat Expression Set 230 1367933_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10833659 - -
          Rat Genome U34 Set Z15123exon#5_s_at - -
          Rat Genome U34 Set rc_AI008131_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21316075 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352980 - -
          RatRef-12 Expression BeadChip ILMN_61942 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline