Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01408490_m1
          See other PIP5K1C GE Assays ›
          Gene Symbol:
          Pip5k1c
          Gene Name:
          phosphatidylinositol-4-phosphate 5-kinase, type I, gamma
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 11267207 - 11294292 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01408490_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pip5k1c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009967.2 NP_001009967.2 15 - 16 1839 61
          NM_001033970.1 NP_001029142.1 15 - 16 1839 61
          XM_006240935.2 XP_006240997.1 15 - 16 1882 61
          XM_008765101.1 XP_008763323.1 15 - 16 1882 61
          XM_008765102.1 XP_008763324.1 15 - 16 1876 61
          XM_008765103.1 XP_008763325.1 15 - 16 1882 61
          GenBank mRNA AY850259.1 - 15 - 16 1778 61
          BC101909.1 - 15 - 16 1811 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700004049.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pip5k1c

          Gene Ontology Categories:

          Function(s) Process(es)

          exocytosis
          phagocytosis
          chemotaxis
          cell adhesion
          axonogenesis
          phosphatidylinositol metabolic process
          phosphatidylinositol phosphate biosynthetic process
          platelet aggregation
          ATP binding
          1-phosphatidylinositol-4-phosphate 5-kinase activity
          talin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P138599 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P138604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31838 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21177 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57961 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148729 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118048 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P138599 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P138604 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P138599 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P138604 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P138604 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P138599 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40279 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47604 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68620 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87160 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10900238 - -
          Affymetrix Rat Gene 1.0 ST Array 10900233 - -
          Affymetrix Rat Gene 1.0 ST Array 10900239 - -
          Affymetrix Rat Gene 1.0 ST Array 10900232 - -
          Affymetrix Rat Gene 1.0 ST Array 10900242 - -
          Affymetrix Rat Gene 1.0 ST Array 10900236 - -
          Affymetrix Rat Gene 1.0 ST Array 10900243 - -
          Affymetrix Rat Gene 1.0 ST Array 10900237 - -
          Affymetrix Rat Gene 1.0 ST Array 10900235 - -
          Affymetrix Rat Gene 1.0 ST Array 10900247 - -
          Affymetrix Rat Gene 1.0 ST Array 10900248 - -
          Affymetrix Rat Gene 1.0 ST Array 10900244 - -
          Affymetrix Rat Gene 1.0 ST Array 10900230 - -
          Affymetrix Rat Gene 1.0 ST Array 10900231 - -
          Affymetrix Rat Gene 1.0 ST Array 10900246 - -
          Affymetrix Rat Gene 1.0 ST Array 10900234 - -
          Affymetrix Rat Gene 1.0 ST Array 10900241 - -
          Affymetrix Rat Gene 1.0 ST Array 10900249 - -
          Affymetrix Rat Gene 1.0 ST Array 10900245 - -
          Affymetrix Rat Gene 1.0 ST Array 10900240 - -
          Affymetrix Rat Gene 1.0 ST Array 10900229 - -
          Affymetrix Rat Gene 1.1 ST Array 10900228 - -
          Rat Genome 230 2.0 Array 1372861_PM_at - -
          Rat Genome 230 2.0 Array 1384016_PM_at - -
          Rat Genome 230 2.0 Array 1384016_at - -
          Rat Genome 230 2.0 Array 1372861_at - -
          Affymetrix HT Rat Focus Array 1372861_at - -
          Affymetrix HT Rat Focus Array 1372861_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700033218.rn.1 TC0700004049.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700033217.rn.1 TC0700004049.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700033216.rn.1 TC0700004049.rn.1 RTA TC
          Rat Expression Set 230 1372861_PM_at - -
          Rat Expression Set 230 1372861_at - -
          Affymetrix Rat Gene 1.0 ST Array 10900228 - -
          Rat Genome U34 Set rc_AA799473_at - -
          Rat Genome U34 Set AA942675_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21984746 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365757 - -
          RatRef-12 Expression BeadChip ILMN_50871 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0