Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01410832_g1
          See other PITX3 GE Assays ›
          Gene Symbol:
          Pitx3
          Gene Name:
          paired-like homeodomain 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 265886766 - 265899947 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01410832_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pitx3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019247.1 NP_062120.1 2 - 3 181 107
          XM_006231478.2 XP_006231540.1 2 - 3 360 107
          GenBank mRNA AJ011005.2 - 2 - 3 181 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007809.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pitx3

          Gene Ontology Categories:

          Function(s) Process(es)

          lens development in camera-type eye
          lens morphogenesis in camera-type eye
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription by RNA polymerase II
          locomotory behavior
          regulation of gene expression
          midbrain development
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          neuron development
          lens fiber cell differentiation
          dopaminergic neuron differentiation
          RNA polymerase II cis-regulatory region sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P072653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151941 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117784 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P072653 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P072653 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P072653 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P758492 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700535127 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33317 - -
          Agilent Rat cDNA Microarray (G4105A) 700535127 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10730535 - -
          Affymetrix Rat Gene 1.0 ST Array 10730536 - -
          Affymetrix Rat Gene 1.0 ST Array 10730538 - -
          Affymetrix Rat Gene 1.0 ST Array 10730537 - -
          Affymetrix Rat Gene 1.1 ST Array 10730534 - -
          Rat Genome 230 2.0 Array 1370222_at - -
          Rat Genome 230 2.0 Array 1370222_PM_at - -
          Affymetrix HT Rat Focus Array 1370222_PM_at - -
          Affymetrix HT Rat Focus Array 1370222_at - -
          Affymetrix Rat Clariom S Assay PSR0100063019.rn.1 TC0100007809.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100063021.rn.1 TC0100007809.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100063022.rn.1 TC0100007809.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100063020.rn.1 TC0100007809.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10730534 - -
          Rat Genome U34 Set AJ011005_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22275333 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365753 - -
          RatRef-12 Expression BeadChip ILMN_66663 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0