Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01411262_m1
          See other PYROXD2 GE Assays ›
          Gene Symbol:
          Pyroxd2
          Gene Name:
          pyridine nucleotide-disulphide oxidoreductase domain 2
          Gene Aliases:
          RGD1303232
          Chromosome Location:
          Chr.1: 261961164 - 261987033 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01411262_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pyroxd2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004261.1 NP_001004261.1 13 - 14 1451 131
          XM_006231394.2 XP_006231456.1 13 - 14 1722 131
          XM_006231396.2 XP_006231458.1 13 - 14 1723 131
          XM_008760424.1 XP_008758646.1 13 - 14 1725 131
          XM_008760425.1 XP_008758647.1 13 - 14 1488 131
          GenBank mRNA BC079368.1 - 13 - 14 1451 131

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007725.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pyroxd2

          Gene Ontology Categories:

          Process(es)

          oxidoreductase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1056401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651552 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11494 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118599 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162359 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139233 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1056401 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1056401 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1056401 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1056401 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68201 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95484 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49886 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10715405 - -
          Affymetrix Rat Gene 1.0 ST Array 10715415 - -
          Affymetrix Rat Gene 1.0 ST Array 10715404 - -
          Affymetrix Rat Gene 1.0 ST Array 10715410 - -
          Affymetrix Rat Gene 1.0 ST Array 10715413 - -
          Affymetrix Rat Gene 1.0 ST Array 10715414 - -
          Affymetrix Rat Gene 1.0 ST Array 10715406 - -
          Affymetrix Rat Gene 1.0 ST Array 10715407 - -
          Affymetrix Rat Gene 1.0 ST Array 10715403 - -
          Affymetrix Rat Gene 1.0 ST Array 10715408 - -
          Affymetrix Rat Gene 1.0 ST Array 10715411 - -
          Affymetrix Rat Gene 1.0 ST Array 10715402 - -
          Affymetrix Rat Gene 1.0 ST Array 10715412 - -
          Affymetrix Rat Gene 1.0 ST Array 10715401 - -
          Affymetrix Rat Gene 1.0 ST Array 10715409 - -
          Affymetrix Rat Gene 1.1 ST Array 10715400 - -
          Rat Genome 230 2.0 Array 1398031_at - -
          Rat Genome 230 2.0 Array 1378802_PM_at - -
          Rat Genome 230 2.0 Array 1398031_PM_at - -
          Rat Genome 230 2.0 Array 1378802_at - -
          Affymetrix HT Rat Focus Array 1398031_PM_at - -
          Affymetrix HT Rat Focus Array 1398031_at - -
          Affymetrix HT Rat Focus Array 1378802_PM_at - -
          Affymetrix HT Rat Focus Array 1378802_at - -
          Affymetrix Rat Clariom S Assay PSR0100062185.rn.1 TC0100007725.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100062189.rn.1 TC0100007725.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100062181.rn.1 TC0100007725.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100062186.rn.1 TC0100007725.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100062176.rn.1 TC0100007725.rn.1 RTA TC
          Rat Expression Set 230 1378802_at - -
          Rat Expression Set 230 1398031_PM_at - -
          Rat Expression Set 230 1398031_at - -
          Rat Expression Set 230 1378802_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10715400 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21520252 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362634 - -
          RatRef-12 Expression BeadChip ILMN_51302 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0