Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01411959_m1
          See other CR1L GE Assays ›
          Gene Symbol:
          Cr1l
          Gene Name:
          complement component (3b/4b) receptor 1-like
          Gene Aliases:
          Cr1, Crry
          Chromosome Location:
          Chr.13: 113824886 - 113872097 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01411959_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cr1l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001005265.1 NP_001005265.1 11 - 12 1707 83
          NM_001005330.1 NP_001005330.1 2079 83
          NM_019301.2 NP_062174.1 12 - 13 1893 83
          GenBank mRNA BC061736.1 - 12 - 13 1662 83
          D42114.1 - 11 - 12 1516 83
          FQ220559.1 - 8 - 9 912 83
          L36532.1 - 12 - 13 1661 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300002267.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cr1l

          Gene Ontology Categories:

          Function(s)

          in utero embryonic development
          complement activation, classical pathway
          multicellular organism development
          female pregnancy
          regulation of complement activation
          innate immune response
          negative regulation of complement activation

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P098779 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321281 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151629 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16864 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161507 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108997 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26688 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35729 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12012 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P098779 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P098779 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12012 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12012 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P098779 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12012 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P132822 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P558888 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88980 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103180 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96152 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10770853 - -
          Affymetrix Rat Gene 1.0 ST Array 10770852 - -
          Affymetrix Rat Gene 1.0 ST Array 10770848 - -
          Affymetrix Rat Gene 1.0 ST Array 10770856 - -
          Affymetrix Rat Gene 1.0 ST Array 10770845 - -
          Affymetrix Rat Gene 1.0 ST Array 10770855 - -
          Affymetrix Rat Gene 1.0 ST Array 10770847 - -
          Affymetrix Rat Gene 1.0 ST Array 10770851 - -
          Affymetrix Rat Gene 1.0 ST Array 10770854 - -
          Affymetrix Rat Gene 1.0 ST Array 10770849 - -
          Affymetrix Rat Gene 1.0 ST Array 10770850 - -
          Affymetrix Rat Gene 1.0 ST Array 10770846 - -
          Affymetrix Rat Gene 1.0 ST Array 10770857 - -
          Affymetrix Rat Gene 1.1 ST Array 10770844 - -
          Rat Genome 230 2.0 Array 1387798_PM_a_at - -
          Rat Genome 230 2.0 Array 1387798_a_at - -
          Rat Genome 230 2.0 Array 1394096_at - -
          Rat Genome 230 2.0 Array 1394096_PM_at - -
          Affymetrix HT Rat Focus Array 1387798_a_at - -
          Affymetrix HT Rat Focus Array 1387798_PM_a_at - -
          Affymetrix HT Rat Focus Array 1394096_at - -
          Affymetrix HT Rat Focus Array 1394096_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300016150.rn.1 TC1300002267.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300016149.rn.1 TC1300002267.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300016146.rn.1 TC1300002267.rn.1 RTA TC
          Rat Expression Set 230 1387798_PM_a_at - -
          Rat Expression Set 230 1387798_a_at - -
          Rat Expression Set 230 1394096_PM_at - -
          Rat Expression Set 230 1394096_at - -
          Affymetrix Rat Gene 1.0 ST Array 10770844 - -
          Rat Genome U34 Set D42116_s_at - -
          Rat Genome U34 Set L36532_s_at - -
          Rat Genome U34 Set rc_AI178368_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21972332 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376671 - -
          RatRef-12 Expression BeadChip ILMN_48169 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0