Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01412047_g1
          See other NEK2L1 GE Assays ›
          Gene Symbol:
          Nek2l1 Nek2
          Gene Name:
          NIMA (never in mitosis gene a)-related kinase 2-like 1
          NIMA-related kinase 2
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.8: 15940935 - 15942589 on Build Rnor_6.0
          Chromosome Location:
          Chr.13: 110511546 - 110524750 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01412047_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nek2l1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_345900.7 XP_345901.4 1 1158 103
          Gene Symbol: Nek2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053691.2 NP_446143.1 7 - 8 1234 103
          GenBank mRNA AF352021.1 - 7 - 8 1103 103

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic sister chromatid segregation
          blastocyst development
          protein phosphorylation
          microtubule-based process
          chromosome segregation
          negative regulation of DNA binding
          protein autophosphorylation
          centrosome separation
          regulation of attachment of spindle microtubules to kinetochore
          mitotic spindle assembly
          negative regulation of centriole-centriole cohesion
          protein serine/threonine kinase activity
          ATP binding
          protein phosphatase binding
          protein kinase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P139013 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P015079 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P015079 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P139013 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P139013 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P015079 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P015079 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P139013 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P16153 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10766770 - -
          Affymetrix Rat Gene 1.0 ST Array 10766771 - -
          Affymetrix Rat Gene 1.0 ST Array 10766767 - -
          Affymetrix Rat Gene 1.0 ST Array 10766766 - -
          Affymetrix Rat Gene 1.0 ST Array 10766768 - -
          Affymetrix Rat Gene 1.0 ST Array 10766773 - -
          Affymetrix Rat Gene 1.0 ST Array 10766772 - -
          Affymetrix Rat Gene 1.0 ST Array 10766769 - -
          Affymetrix Rat Gene 1.0 ST Array 10908128 - -
          Affymetrix Rat Gene 1.1 ST Array 10908127 - -
          Affymetrix Rat Gene 1.1 ST Array 10766765 - -
          Affymetrix Rat Clariom S Assay PSR1300007572.rn.1 TC1300001068.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300007566.rn.1 TC1300001068.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300007568.rn.1 TC1300001068.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300007571.rn.1 TC1300001068.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10908127 - -
          Affymetrix Rat Gene 1.0 ST Array 10766765 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20722922 - -
          Array Name Probe IDs Targets Target Type
          RatRef-12 Expression BeadChip ILMN_67159 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0