Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01412174_m1
          See other GREM2 GE Assays ›
          Gene Symbol:
          Grem2
          Gene Name:
          gremlin 2
          Gene Aliases:
          RGD1560008
          Chromosome Location:
          Chr.13: 92894431 - 92988137 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01412174_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Grem2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105974.1 NP_001099444.1 1 - 2 240 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001986.rn.1

          Additional Information:

          This assay is in the 5'UTR region of the NM_001105974.1 transcripts.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          embryonic body morphogenesis
          cytokine-mediated signaling pathway
          negative regulation of BMP signaling pathway
          sequestering of BMP from receptor via BMP binding
          determination of dorsal identity
          regulation of cytokine activity
          cytokine activity
          heparin binding
          BMP binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P162975 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695798 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139905 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19803 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86563 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167717 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99563 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94014 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109575 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P162975 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13931 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P162975 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13931 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P162975 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13931 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P395985 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15779 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37175 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10770118 - -
          Affymetrix Rat Gene 1.1 ST Array 10770117 - -
          Rat Genome 230 2.0 Array 1378163_at - -
          Rat Genome 230 2.0 Array 1378163_PM_at - -
          Rat Genome 230 2.0 Array 1380799_at - -
          Rat Genome 230 2.0 Array 1380799_PM_at - -
          Affymetrix HT Rat Focus Array 1378163_at - -
          Affymetrix HT Rat Focus Array 1378163_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300014225.rn.1 TC1300001986.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300014223.rn.1 TC1300001986.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300014224.rn.1 TC1300001986.rn.1 RTA TC
          Rat Expression Set 230 1380799_PM_at - -
          Rat Expression Set 230 1378163_at - -
          Rat Expression Set 230 1380799_at - -
          Rat Expression Set 230 1378163_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10770117 - -
          Rat Genome U34 Set rc_AA817956_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22288019 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358228 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-66756dbf47-27kf9:80/100.66.130.62:80.
          git-commit: ed967532dff6834307dac93af002e5c8e22ba7c9
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline