Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01412613_g1
          See other RPS21 GE Assays ›
          Gene Symbol:
          Rps21
          Gene Name:
          ribosomal protein S21
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 175629087 - 175630205 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01412613_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rps21
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031111.1 NP_112373.1 5 - 6 280 90
          XM_006235823.1 XP_006235885.1 5 - 6 369 90
          GenBank mRNA BC058464.1 - 5 - 6 285 90
          FQ210654.1 - 5 - 6 283 90
          FQ210736.1 - 5 - 6 283 90
          FQ210823.1 - 5 - 6 281 90
          FQ210868.1 - 5 - 6 282 90
          FQ212305.1 - 5 - 6 282 90
          FQ212673.1 - 5 - 6 275 90
          FQ217300.1 - 5 - 6 281 90
          FQ221031.1 - 5 - 6 283 90
          FQ221222.1 - 5 - 6 283 90
          FQ221376.1 - 5 - 6 281 90
          FQ221504.1 - 5 - 6 281 90
          FQ221719.1 - 5 - 6 282 90
          FQ221773.1 - 5 - 6 281 90
          FQ221882.1 - 5 - 6 281 90
          FQ221895.1 - 5 - 6 281 90
          FQ222195.1 - 5 - 6 281 90
          FQ222197.1 - 5 - 6 90
          FQ222346.1 - 5 - 6 282 90
          FQ222395.1 - 5 - 6 281 90
          FQ222640.1 - 5 - 6 284 90
          FQ222661.1 - 5 - 6 281 90
          FQ222692.1 - 5 - 6 283 90
          FQ222717.1 - 5 - 6 281 90
          FQ222985.1 - 5 - 6 281 90
          FQ223083.1 - 5 - 6 281 90
          FQ223085.1 - 5 - 6 281 90
          FQ223126.1 - 5 - 6 281 90
          FQ223140.1 - 5 - 6 280 90
          FQ223324.1 - 5 - 6 281 90
          FQ223458.1 - 5 - 6 282 90
          FQ224512.1 - 5 - 6 282 90
          FQ224728.1 - 5 - 6 283 90
          FQ228462.1 - 5 - 6 281 90
          FQ228580.1 - 5 - 6 282 90
          FQ228593.1 - 5 - 6 281 90
          FQ228604.1 - 5 - 6 286 90
          FQ229072.1 - 5 - 6 276 90
          FQ229301.1 - 5 - 6 285 90
          FQ229813.1 - 5 - 6 281 90
          X79059.1 - 5 - 6 280 90

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rps21

          Gene Ontology Categories:

          Function(s) Process(es)

          endonucleolytic cleavage in ITS1 to separate SSU-rRNA from 5.8S rRNA and LSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
          endonucleolytic cleavage to generate mature 3'-end of SSU-rRNA from (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
          translational elongation
          biological_process
          ribosomal small subunit biogenesis
          molecular_function
          RNA binding
          structural constituent of ribosome
          ribosome binding
          protein N-terminus binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P310152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511878 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P310152 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P310152 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P310152 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P310152 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700484168 - -
          Agilent Rat cDNA Microarray (G4105A) 700484168 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10842806 - -
          Affymetrix Rat Gene 1.0 ST Array 10842807 - -
          Affymetrix Rat Gene 1.0 ST Array 10842810 - -
          Affymetrix Rat Gene 1.0 ST Array 10873651 - -
          Affymetrix Rat Gene 1.0 ST Array 10842809 - -
          Affymetrix Rat Gene 1.0 ST Array 10909760 - -
          Affymetrix Rat Gene 1.0 ST Array 10909759 - -
          Affymetrix Rat Gene 1.0 ST Array 10842808 - -
          Affymetrix Rat Gene 1.0 ST Array 10873650 - -
          Affymetrix Rat Gene 1.0 ST Array 10938963 - -
          Affymetrix Rat Gene 1.1 ST Array 10873649 - -
          Affymetrix Rat Gene 1.1 ST Array 10909758 - -
          Affymetrix Rat Gene 1.1 ST Array 10938962 - -
          Affymetrix Rat Gene 1.1 ST Array 10842805 - -
          Rat Genome 230 2.0 Array 1398852_PM_at - -
          Rat Genome 230 2.0 Array 1398852_at - -
          Affymetrix HT Rat Focus Array 1398852_PM_at - -
          Affymetrix HT Rat Focus Array 1398852_at - -
          Affymetrix Rat Clariom S Assay PSR0300018855.rn.1 TC0300002560.rn.1 RTA TC
          Rat Expression Set 230 1398852_at - -
          Rat Expression Set 230 1398852_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10873649 - -
          Affymetrix Rat Gene 1.0 ST Array 10842805 - -
          Affymetrix Rat Gene 1.0 ST Array 10909758 - -
          Affymetrix Rat Gene 1.0 ST Array 10938962 - -
          Rat Genome U34 Set rc_AI178207_at - -
          Rat Genome U34 Set rc_AA955374_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21653893 - -
          ABI Rat Genome Survey Microarray 21983018 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358719 - -
          RatRef-12 Expression BeadChip ILMN_57081 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-ztd5s:80/100.66.134.6:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0