Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01412930_m1
          See other HSPH1 GE Assays ›
          Gene Symbol:
          Hsph1
          Gene Name:
          heat shock 105/110 protein 1
          Gene Aliases:
          Hsp105
          Chromosome Location:
          Chr.12: 6322685 - 6341898 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01412930_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hsph1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011901.1 NP_001011901.1 17 - 18 2564 127
          XM_008768947.1 XP_008767169.1 16 - 17 2325 127
          GenBank mRNA BC081945.1 - 17 - 18 2564 127

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000977.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hsph1

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of transcription by RNA polymerase II
          chaperone cofactor-dependent protein refolding
          positive regulation of protein tyrosine kinase activity
          negative regulation of establishment of protein localization to mitochondrion
          negative regulation of intrinsic apoptotic signaling pathway in response to hydrogen peroxide
          negative regulation of p38MAPK cascade
          negative regulation of apoptotic signaling pathway
          ATP binding
          alpha-tubulin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P121374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963623 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169475 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131450 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P121374 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P121374 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P121374 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P121374 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43854 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69472 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10756357 - -
          Affymetrix Rat Gene 1.0 ST Array 10756353 - -
          Affymetrix Rat Gene 1.0 ST Array 10756349 - -
          Affymetrix Rat Gene 1.0 ST Array 10756345 - -
          Affymetrix Rat Gene 1.0 ST Array 10756352 - -
          Affymetrix Rat Gene 1.0 ST Array 10756356 - -
          Affymetrix Rat Gene 1.0 ST Array 10756360 - -
          Affymetrix Rat Gene 1.0 ST Array 10756344 - -
          Affymetrix Rat Gene 1.0 ST Array 10756358 - -
          Affymetrix Rat Gene 1.0 ST Array 10756359 - -
          Affymetrix Rat Gene 1.0 ST Array 10756361 - -
          Affymetrix Rat Gene 1.0 ST Array 10756348 - -
          Affymetrix Rat Gene 1.0 ST Array 10756362 - -
          Affymetrix Rat Gene 1.0 ST Array 10756351 - -
          Affymetrix Rat Gene 1.0 ST Array 10756355 - -
          Affymetrix Rat Gene 1.0 ST Array 10756354 - -
          Affymetrix Rat Gene 1.0 ST Array 10756350 - -
          Affymetrix Rat Gene 1.1 ST Array 10756343 - -
          Rat Genome 230 2.0 Array 1385620_at - -
          Rat Genome 230 2.0 Array 1385620_PM_at - -
          Rat Genome 230 2.0 Array 1388898_PM_at - -
          Rat Genome 230 2.0 Array 1388898_at - -
          Affymetrix HT Rat Focus Array 1385620_PM_at - -
          Affymetrix HT Rat Focus Array 1388898_at - -
          Affymetrix HT Rat Focus Array 1385620_at - -
          Affymetrix HT Rat Focus Array 1388898_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200008542.rn.1 TC1200000977.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200008543.rn.1 TC1200000977.rn.1 RTA TC
          Rat Expression Set 230 1388898_PM_at - -
          Rat Expression Set 230 1388898_at - -
          Rat Expression Set 230 1385620_PM_at - -
          Rat Expression Set 230 1385620_at - -
          Affymetrix Rat Gene 1.0 ST Array 10756343 - -
          Rat Genome U34 Set rc_AI236601_at - -
          Rat Genome U34 Set AA108277_at - -
          Rat Genome U34 Set rc_AI236601_at - -
          Rat Genome U34 Set AA108277_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21832208 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1530359 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375803 - -
          RatRef-12 Expression BeadChip ILMN_161583 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0