Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01413425_m1
          See other NOA1 GE Assays ›
          Gene Symbol:
          Noa1
          Gene Name:
          nitric oxide associated 1
          Gene Aliases:
          RGD1359460
          Chromosome Location:
          Chr.14: 33107825 - 33124172 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01413425_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Noa1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001006959.1 NP_001006960.1 4 - 5 1732 68
          XM_006250845.2 XP_006250907.1 4 - 5 2047 68
          GenBank mRNA BC083902.1 - 4 - 5 1732 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400000370.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Noa1

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell death
          mitochondrial translation
          ribosome biogenesis
          regulation of cellular respiration
          RNA binding
          GTPase activity
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P991045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225505 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35786 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62800 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P991045 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P991045 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P991045 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P991045 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54110 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21120 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10772184 - -
          Affymetrix Rat Gene 1.0 ST Array 10772180 - -
          Affymetrix Rat Gene 1.0 ST Array 10772181 - -
          Affymetrix Rat Gene 1.0 ST Array 10772177 - -
          Affymetrix Rat Gene 1.0 ST Array 10772178 - -
          Affymetrix Rat Gene 1.0 ST Array 10772179 - -
          Affymetrix Rat Gene 1.0 ST Array 10772182 - -
          Affymetrix Rat Gene 1.0 ST Array 10772185 - -
          Affymetrix Rat Gene 1.1 ST Array 10772176 - -
          Rat Genome 230 2.0 Array 1372804_at - -
          Rat Genome 230 2.0 Array 1372804_PM_at - -
          Affymetrix HT Rat Focus Array 1372804_at - -
          Affymetrix HT Rat Focus Array 1372804_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1400002929.rn.1 TC1400000370.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400002930.rn.1 TC1400000370.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400002927.rn.1 TC1400000370.rn.1 RTA TC
          Rat Expression Set 230 1372804_at - -
          Rat Expression Set 230 1372804_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10772176 - -
          Rat Genome U34 Set rc_AI175555_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21392804 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368907 - -
          RatRef-12 Expression BeadChip ILMN_53028 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0