Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01415151_m1
          See other LOC103689999 GE Assays ›
          Gene Symbol:
          LOC103689999 Sccpdh
          Gene Name:
          saccharopine dehydrogenase-like oxidoreductase
          saccharopine dehydrogenase (putative)
          Gene Aliases:
          -
          RGD1311440
          Chromosome Location:
          Chr.13: 97701878 - 97724165 on Build Rnor_6.0
          Chromosome Location:
          Chr.13: 97941720 - 97963474 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01415151_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC103689999
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008769879.1 XP_008768101.1 11 - 12 1478 57
          Gene Symbol: Sccpdh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013985.1 NP_001014007.1 11 - 12 1296 57
          GenBank mRNA BC061579.1 - 1100 57
          BC079215.1 - 11 - 12 1296 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000891.rn.1, TC1300000895.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC103689999
          Click here to find siRNAs for Gene Sccpdh

          Gene Ontology Categories:

          Process(es)

          oxidoreductase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P224789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306927 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63778 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87423 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6578 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P224789 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P224789 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P224789 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P224789 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P224793 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16802 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64258 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10766118 - -
          Affymetrix Rat Gene 1.0 ST Array 10766107 - -
          Affymetrix Rat Gene 1.0 ST Array 10766109 - -
          Affymetrix Rat Gene 1.0 ST Array 10766112 - -
          Affymetrix Rat Gene 1.0 ST Array 10766114 - -
          Affymetrix Rat Gene 1.0 ST Array 10766106 - -
          Affymetrix Rat Gene 1.0 ST Array 10766116 - -
          Affymetrix Rat Gene 1.0 ST Array 10766119 - -
          Affymetrix Rat Gene 1.0 ST Array 10766117 - -
          Affymetrix Rat Gene 1.0 ST Array 10766113 - -
          Affymetrix Rat Gene 1.0 ST Array 10766110 - -
          Affymetrix Rat Gene 1.0 ST Array 10766108 - -
          Affymetrix Rat Gene 1.0 ST Array 10766115 - -
          Affymetrix Rat Gene 1.0 ST Array 10766111 - -
          Affymetrix Rat Gene 1.1 ST Array 10766105 - -
          Rat Genome 230 2.0 Array 1393264_at - -
          Rat Genome 230 2.0 Array 1373652_at - -
          Rat Genome 230 2.0 Array 1373652_PM_at - -
          Rat Genome 230 2.0 Array 1372741_PM_at - -
          Rat Genome 230 2.0 Array 1372741_at - -
          Rat Genome 230 2.0 Array 1393264_PM_at - -
          Affymetrix HT Rat Focus Array 1372741_at - -
          Affymetrix HT Rat Focus Array 1372741_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300006276.rn.1 TC1300000895.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300006220.rn.1 TC1300000891.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300006221.rn.1 TC1300000891.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300006275.rn.1 TC1300000895.rn.1 RTA TC
          Rat Expression Set 230 1372741_PM_at - -
          Rat Expression Set 230 1372741_at - -
          Affymetrix Rat Gene 1.0 ST Array 10766105 - -
          Rat Genome U34 Set rc_AA850814_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21381088 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349324 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0