Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01415566_m1
          See other DTD1 GE Assays ›
          Gene Symbol:
          Dtd1
          Gene Name:
          D-tyrosyl-tRNA deacylase 1
          Gene Aliases:
          Hars2
          Chromosome Location:
          Chr.3: 138770575 - 138931544 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01415566_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dtd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108594.1 NP_001102064.1 4 - 5 541 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300001890.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA metabolic process
          regulation of translational fidelity
          D-amino acid catabolic process
          aminoacyl-tRNA editing activity
          aminoacyl-tRNA ligase activity
          D-tyrosyl-tRNA(Tyr) deacylase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P410192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317581 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94627 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84221 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15840 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175642 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18826 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167070 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130970 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126253 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52117 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180579 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157615 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53697 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151035 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76758 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46277 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10885 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121613 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52641 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129044 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24731 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P410192 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P410192 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P410192 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P410192 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29139 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53719 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9399 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97753 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10840424 - -
          Affymetrix Rat Gene 1.0 ST Array 10840421 - -
          Affymetrix Rat Gene 1.0 ST Array 10840420 - -
          Affymetrix Rat Gene 1.0 ST Array 10840423 - -
          Affymetrix Rat Gene 1.0 ST Array 10840419 - -
          Affymetrix Rat Gene 1.0 ST Array 10840422 - -
          Affymetrix Rat Gene 1.1 ST Array 10840418 - -
          Rat Genome 230 2.0 Array 1388428_at - -
          Rat Genome 230 2.0 Array 1388428_PM_at - -
          Affymetrix HT Rat Focus Array 1388428_at - -
          Affymetrix HT Rat Focus Array 1388428_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300013562.rn.1 TC0300001890.rn.1 RTA TC
          Rat Expression Set 230 1388428_at - -
          Rat Expression Set 230 1388428_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10840418 - -
          Rat Genome U34 Set rc_AI233365_at - -
          Rat Genome U34 Set rc_AI233365_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21574106 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364969 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0