Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01415611_m1
          See other XRN2 GE Assays ›
          Gene Symbol:
          Xrn2
          Gene Name:
          5'-3' exoribonuclease 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 141224855 - 141298615 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01415611_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Xrn2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108596.1 NP_001102066.1 15 - 16 1666 65
          XM_006235156.2 XP_006235218.1 16 - 17 2065 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300001915.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA catabolic process, exonucleolytic
          spermatogenesis
          biological_process
          cell growth
          RNA metabolic process
          transcription termination site sequence-specific DNA binding
          molecular_function
          RNA binding
          5'-3' exonuclease activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P003744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577092 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137603 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111047 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10844 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154638 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125288 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136565 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63706 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156405 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107004 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P003744 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P003744 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P003744 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1026431 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P575227 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P489956 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16267 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50546 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10840545 - -
          Affymetrix Rat Gene 1.0 ST Array 10840544 - -
          Affymetrix Rat Gene 1.0 ST Array 10840537 - -
          Affymetrix Rat Gene 1.0 ST Array 10840547 - -
          Affymetrix Rat Gene 1.0 ST Array 10840538 - -
          Affymetrix Rat Gene 1.0 ST Array 10840550 - -
          Affymetrix Rat Gene 1.0 ST Array 10840541 - -
          Affymetrix Rat Gene 1.0 ST Array 10840528 - -
          Affymetrix Rat Gene 1.0 ST Array 10840524 - -
          Affymetrix Rat Gene 1.0 ST Array 10840522 - -
          Affymetrix Rat Gene 1.0 ST Array 10840539 - -
          Affymetrix Rat Gene 1.0 ST Array 10840536 - -
          Affymetrix Rat Gene 1.0 ST Array 10840533 - -
          Affymetrix Rat Gene 1.0 ST Array 10840523 - -
          Affymetrix Rat Gene 1.0 ST Array 10840530 - -
          Affymetrix Rat Gene 1.0 ST Array 10840546 - -
          Affymetrix Rat Gene 1.0 ST Array 10840542 - -
          Affymetrix Rat Gene 1.0 ST Array 10840532 - -
          Affymetrix Rat Gene 1.0 ST Array 10840548 - -
          Affymetrix Rat Gene 1.0 ST Array 10840549 - -
          Affymetrix Rat Gene 1.0 ST Array 10840527 - -
          Affymetrix Rat Gene 1.0 ST Array 10840535 - -
          Affymetrix Rat Gene 1.0 ST Array 10840525 - -
          Affymetrix Rat Gene 1.0 ST Array 10840531 - -
          Affymetrix Rat Gene 1.0 ST Array 10840529 - -
          Affymetrix Rat Gene 1.0 ST Array 10840534 - -
          Affymetrix Rat Gene 1.0 ST Array 10840540 - -
          Affymetrix Rat Gene 1.0 ST Array 10840543 - -
          Affymetrix Rat Gene 1.0 ST Array 10840526 - -
          Affymetrix Rat Gene 1.1 ST Array 10840519 - -
          Rat Genome 230 2.0 Array 1393702_at - -
          Rat Genome 230 2.0 Array 1393702_PM_at - -
          Rat Genome 230 2.0 Array 1391412_PM_at - -
          Rat Genome 230 2.0 Array 1391412_at - -
          Affymetrix HT Rat Focus Array 1391412_at - -
          Affymetrix HT Rat Focus Array 1393702_PM_at - -
          Affymetrix HT Rat Focus Array 1393702_at - -
          Affymetrix HT Rat Focus Array 1391412_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300013829.rn.1 TC0300001915.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300013828.rn.1 TC0300001915.rn.1 RTA TC
          Rat Expression Set 230 1391412_PM_at - -
          Rat Expression Set 230 1391412_at - -
          Rat Expression Set 230 1393702_PM_at - -
          Rat Expression Set 230 1393702_at - -
          Affymetrix Rat Gene 1.0 ST Array 10840519 - -
          Rat Genome U34 Set rc_AA891938_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22178479 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361377 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0