Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01415707_g1
          See other GZF1 GE Assays ›
          Gene Symbol:
          Gzf1
          Gene Name:
          GDNF-inducible zinc finger protein 1
          Gene Aliases:
          RGD1562321, Zfp336
          Chromosome Location:
          Chr.3: 143003678 - 143015906 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01415707_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gzf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107788.1 NP_001101258.1 5 - 6 2049 102
          XM_006235153.2 XP_006235215.1 5 - 6 1916 102
          XM_006235154.2 XP_006235216.1 5 - 6 3028 102
          XM_008762274.1 XP_008760496.1 6 - 7 2207 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300001942.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          branching involved in ureteric bud morphogenesis
          transcription, DNA-templated
          negative regulation of transcription, DNA-templated
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          sequence-specific DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P125370 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P19499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744243 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46400 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P19499 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P125370 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P125370 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P19499 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P19499 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P125370 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P19366 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P19499 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10840572 - -
          Affymetrix Rat Gene 1.0 ST Array 10840573 - -
          Affymetrix Rat Gene 1.0 ST Array 10840574 - -
          Affymetrix Rat Gene 1.0 ST Array 10840575 - -
          Affymetrix Rat Gene 1.0 ST Array 10840576 - -
          Affymetrix Rat Gene 1.1 ST Array 10840571 - -
          Rat Genome 230 2.0 Array 1376159_PM_at - -
          Rat Genome 230 2.0 Array 1376159_at - -
          Affymetrix HT Rat Focus Array 1376159_PM_at - -
          Affymetrix HT Rat Focus Array 1376159_at - -
          Affymetrix Rat Clariom S Assay PSR0300013946.rn.1 TC0300001942.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300013945.rn.1 TC0300001942.rn.1 RTA TC
          Rat Expression Set 230 1376159_at - -
          Rat Expression Set 230 1376159_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10840571 - -
          Rat Genome U34 Set rc_AI177089_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21232447 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349321 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0