Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01416343_m1
          See other ZRSR2 GE Assays ›
          Gene Symbol:
          Zrsr2
          Gene Name:
          zinc finger (CCCH type), RNA binding motif and serine/arginine rich 2
          Gene Aliases:
          RGD1559763
          Chromosome Location:
          Chr.X: 32304720 - 32327419 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01416343_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zrsr2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006256887.2 XP_006256949.1 11 - 12 1191 72
          XM_217612.5 XP_217612.5 10 - 11 1314 72

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          spliceosomal complex assembly
          mRNA splicing, via spliceosome
          nucleotide binding
          RNA binding
          pre-mRNA 3'-splice site binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P622645 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P622645 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P622645 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P622645 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P513911 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P622645 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10933459 - -
          Affymetrix Rat Gene 1.0 ST Array 10933462 - -
          Affymetrix Rat Gene 1.0 ST Array 10933468 - -
          Affymetrix Rat Gene 1.0 ST Array 10933466 - -
          Affymetrix Rat Gene 1.0 ST Array 10933464 - -
          Affymetrix Rat Gene 1.0 ST Array 10933463 - -
          Affymetrix Rat Gene 1.0 ST Array 10933461 - -
          Affymetrix Rat Gene 1.0 ST Array 10933467 - -
          Affymetrix Rat Gene 1.0 ST Array 10933465 - -
          Affymetrix Rat Gene 1.0 ST Array 10933458 - -
          Affymetrix Rat Gene 1.0 ST Array 10933460 - -
          Affymetrix Rat Gene 1.0 ST Array 10933469 - -
          Rat Genome 230 2.0 Array 1395379_at - -
          Rat Genome 230 2.0 Array 1395379_PM_at - -
          Rat Genome 230 2.0 Array 1375058_at - -
          Rat Genome 230 2.0 Array 1375058_PM_at - -
          Affymetrix HT Rat Focus Array 1395379_at - -
          Affymetrix HT Rat Focus Array 1375058_at - -
          Affymetrix HT Rat Focus Array 1375058_PM_at - -
          Affymetrix HT Rat Focus Array 1395379_PM_at - -
          Rat Expression Set 230 1375058_PM_at - -
          Rat Expression Set 230 1375058_at - -
          Rat Expression Set 230 1395379_PM_at - -
          Rat Expression Set 230 1395379_at - -
          Affymetrix Rat Gene 1.0 ST Array 10933457 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350290 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364937 - -
          RatRef-12 Expression BeadChip ILMN_50142 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0