Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01416785_g1
          See other PLK3 GE Assays ›
          Gene Symbol:
          Plk3
          Gene Name:
          polo-like kinase 3
          Gene Aliases:
          Cnk
          Chromosome Location:
          Chr.5: 135997725 - 136002900 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01416785_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Plk3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022187.1 NP_071523.1 10 - 11 1370 63
          GenBank mRNA AF136584.1 - 10 - 11 1166 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500003434.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          negative regulation of transcription by RNA polymerase II
          response to reactive oxygen species
          protein phosphorylation
          apoptotic process
          response to osmotic stress
          cellular response to DNA damage stimulus
          response to radiation
          cytoplasmic microtubule organization
          regulation of cytokinesis
          negative regulation of apoptotic process
          protein kinase B signaling
          mitotic G1/S transition checkpoint signaling
          regulation of synaptic plasticity
          regulation of long-term neuronal synaptic plasticity
          regulation of cell division
          Golgi disassembly
          positive regulation of intracellular protein transport
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process involved in cellular response to hypoxia
          p53 binding
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P135224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250990 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60585 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P135224 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P135224 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P135224 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P543283 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P135224 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700821368 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36337 - -
          Agilent Rat cDNA Microarray (G4105A) 700821368 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10878942 - -
          Affymetrix Rat Gene 1.0 ST Array 10878949 - -
          Affymetrix Rat Gene 1.0 ST Array 10878945 - -
          Affymetrix Rat Gene 1.0 ST Array 10878943 - -
          Affymetrix Rat Gene 1.0 ST Array 10878952 - -
          Affymetrix Rat Gene 1.0 ST Array 10878940 - -
          Affymetrix Rat Gene 1.0 ST Array 10878947 - -
          Affymetrix Rat Gene 1.0 ST Array 10878950 - -
          Affymetrix Rat Gene 1.0 ST Array 10878948 - -
          Affymetrix Rat Gene 1.0 ST Array 10878944 - -
          Affymetrix Rat Gene 1.0 ST Array 10878953 - -
          Affymetrix Rat Gene 1.0 ST Array 10878941 - -
          Affymetrix Rat Gene 1.0 ST Array 10878946 - -
          Affymetrix Rat Gene 1.0 ST Array 10878939 - -
          Affymetrix Rat Gene 1.0 ST Array 10878951 - -
          Affymetrix Rat Gene 1.1 ST Array 10878938 - -
          Rat Genome 230 2.0 Array 1393155_PM_at - -
          Rat Genome 230 2.0 Array 1393155_at - -
          Affymetrix HT Rat Focus Array 1393155_at - -
          Affymetrix HT Rat Focus Array 1393155_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500025075.rn.1 TC0500003434.rn.1 RTA TC
          Rat Expression Set 230 1393155_PM_at - -
          Rat Expression Set 230 1393155_at - -
          Affymetrix Rat Gene 1.0 ST Array 10878938 - -
          Rat Genome U34 Set rc_AA957449_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21427554 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0