Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01418405_m1
          See other REC114 GE Assays ›
          Gene Symbol:
          Rec114
          Gene Name:
          REC114 meiotic recombination protein
          Gene Aliases:
          RGD1311874
          Chromosome Location:
          Chr.8: 63445644 - 63533643 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01418405_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rec114
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106825.1 NP_001100295.1 4 - 5 582 63
          XM_006243157.2 XP_006243219.1 4 - 5 714 63
          XM_006243158.2 XP_006243220.1 4 - 5 727 63
          XM_006243160.2 XP_006243222.1 4 - 5 730 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002659.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P141747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853326 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156196 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165223 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135450 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17226 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74115 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68587 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142600 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89192 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116759 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P141747 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P141747 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P141747 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P568943 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103837 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100172 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56713 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10917919 - -
          Affymetrix Rat Gene 1.0 ST Array 10917922 - -
          Affymetrix Rat Gene 1.0 ST Array 10917918 - -
          Affymetrix Rat Gene 1.0 ST Array 10917921 - -
          Affymetrix Rat Gene 1.0 ST Array 10917917 - -
          Affymetrix Rat Gene 1.0 ST Array 10917923 - -
          Affymetrix Rat Gene 1.1 ST Array 10917916 - -
          Rat Genome 230 2.0 Array 1372387_PM_at - -
          Rat Genome 230 2.0 Array 1372387_at - -
          Rat Genome 230 2.0 Array 1394606_at - -
          Rat Genome 230 2.0 Array 1394606_PM_at - -
          Affymetrix HT Rat Focus Array 1372387_at - -
          Affymetrix HT Rat Focus Array 1372387_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800021614.rn.1 TC0800002659.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800021615.rn.1 TC0800002659.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800021617.rn.1 TC0800002659.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800021616.rn.1 TC0800002659.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800021620.rn.1 TC0800002659.rn.1 RTA TC
          Rat Expression Set 230 1372387_at - -
          Rat Expression Set 230 1372387_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10917916 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20754845 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361339 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0