Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01419872_m1
          See other FKBP8 GE Assays ›
          Gene Symbol:
          Fkbp8
          Gene Name:
          FK506 binding protein 8
          Gene Aliases:
          FKBP-8
          Chromosome Location:
          Chr.16: 20645956 - 20652890 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01419872_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fkbp8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037180.1 NP_001032257.1 5 - 6 814 72
          GenBank mRNA BC107454.1 - 5 - 6 814 72
          FQ216680.1 - 1 - 2 208 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600001400.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fkbp8

          Gene Ontology Categories:

          Function(s) Process(es)

          protein peptidyl-prolyl isomerization
          cell fate specification
          apoptotic process
          smoothened signaling pathway
          dorsal/ventral pattern formation
          regulation of gene expression
          dorsal/ventral neural tube patterning
          neural tube development
          regulation of BMP signaling pathway
          positive regulation of BMP signaling pathway
          multicellular organism growth
          camera-type eye development
          negative regulation of apoptotic process
          chaperone-mediated protein folding
          peptidyl-prolyl cis-trans isomerase activity
          FK506 binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P182792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832771 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P182792 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P182792 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P182792 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P182792 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10790998 - -
          Affymetrix Rat Gene 1.0 ST Array 10790992 - -
          Affymetrix Rat Gene 1.0 ST Array 10790994 - -
          Affymetrix Rat Gene 1.0 ST Array 10790999 - -
          Affymetrix Rat Gene 1.0 ST Array 10790997 - -
          Affymetrix Rat Gene 1.0 ST Array 10790995 - -
          Affymetrix Rat Gene 1.0 ST Array 10790993 - -
          Affymetrix Rat Gene 1.0 ST Array 10790996 - -
          Affymetrix Rat Gene 1.1 ST Array 10790991 - -
          Rat Genome 230 2.0 Array 1371528_PM_at - -
          Rat Genome 230 2.0 Array 1376070_PM_at - -
          Rat Genome 230 2.0 Array 1371528_at - -
          Rat Genome 230 2.0 Array 1376070_at - -
          Affymetrix HT Rat Focus Array 1371528_PM_at - -
          Affymetrix HT Rat Focus Array 1376070_PM_at - -
          Affymetrix HT Rat Focus Array 1376070_at - -
          Affymetrix HT Rat Focus Array 1371528_at - -
          Affymetrix Rat Clariom S Assay PSR1600010709.rn.1 TC1600001400.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600010712.rn.1 TC1600001400.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600010711.rn.1 TC1600001400.rn.1 RTA TC
          Rat Expression Set 230 1376070_PM_at - -
          Rat Expression Set 230 1371528_at - -
          Rat Expression Set 230 1376070_at - -
          Rat Expression Set 230 1371528_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10790991 - -
          Rat Genome U34 Set rc_AI236623_at - -
          Rat Genome U34 Set rc_AI177742_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21643588 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360786 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0