Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01420266_m1
          See other CRTC1 GE Assays ›
          Gene Symbol:
          Crtc1
          Gene Name:
          CREB regulated transcription coactivator 1
          Gene Aliases:
          Mect1, TORC-1, TORC1
          Chromosome Location:
          Chr.16: 20740750 - 20798142 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01420266_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Crtc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001047115.1 NP_001040580.1 9 - 10 1010 64
          XM_006252936.2 XP_006252998.1 9 - 10 1086 64
          XM_008771147.1 XP_008769369.1 9 - 10 1086 64
          GenBank mRNA DQ538521.1 - 9 - 10 1010 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600000302.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          memory
          positive regulation of CREB transcription factor activity
          entrainment of circadian clock by photoperiod
          positive regulation of transcription by RNA polymerase II
          rhythmic process
          protein homotetramerization
          membrane hyperpolarization
          positive regulation of dendrite development
          positive regulation of long-term synaptic potentiation
          molecular_function
          cAMP response element binding protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P111649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566519 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58799 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79070 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50310 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12843 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168210 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P111649 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P111649 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P111649 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88790 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27077 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69186 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104668 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10787576 - -
          Affymetrix Rat Gene 1.0 ST Array 10787578 - -
          Affymetrix Rat Gene 1.0 ST Array 10787574 - -
          Affymetrix Rat Gene 1.0 ST Array 10787572 - -
          Affymetrix Rat Gene 1.0 ST Array 10787573 - -
          Affymetrix Rat Gene 1.0 ST Array 10787571 - -
          Affymetrix Rat Gene 1.0 ST Array 10787577 - -
          Affymetrix Rat Gene 1.0 ST Array 10787575 - -
          Affymetrix Rat Gene 1.1 ST Array 10787570 - -
          Affymetrix Rat Clariom S Assay PSR1600003101.rn.1 TC1600000302.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600003100.rn.1 TC1600000302.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10787570 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21795450 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0