Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01420976_m1
          See other NEWGENE_1562258 GE Assays ›
          Gene Symbol:
          NEWGENE_1562258 Ino80b
          Gene Name:
          INO80 complex subunit B
          INO80 complex subunit B
          Gene Aliases:
          Ino80b
          RGD1562258, Znhit4
          Chromosome Location:
          Chr.4: 114817603 - 114823889 on Build Rnor_6.0
          Chromosome Location:
          Chr.4: 113954612 - 113957774 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01420976_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NEWGENE_1562258
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008763087.1 XP_008761309.1 4 - 5 690 85
          XM_008763088.1 XP_008761310.1 4 - 5 690 85
          XM_008763089.1 XP_008761311.1 4 - 5 928 85
          Gene Symbol: Ino80b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001048233.1 NP_001041698.1 541 85
          GenBank mRNA BC105625.1 - 541 85
          FQ214280.1 - 928 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003175.rn.1, TC0400004158.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin remodeling
          regulation of transcription, DNA-templated
          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1023611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163666 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1023611 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1023611 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1023611 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1023611 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10863500 - -
          Affymetrix Rat Gene 1.0 ST Array 10863499 - -
          Affymetrix Rat Gene 1.0 ST Array 10863501 - -
          Affymetrix Rat Gene 1.0 ST Array 10863497 - -
          Affymetrix Rat Gene 1.0 ST Array 10863498 - -
          Affymetrix Rat Gene 1.1 ST Array 10863496 - -
          Rat Genome 230 2.0 Array 1374450_at - -
          Rat Genome 230 2.0 Array 1374450_PM_at - -
          Affymetrix HT Rat Focus Array 1374450_at - -
          Affymetrix HT Rat Focus Array 1374450_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400022472.rn.1 TC0400003175.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400029847.rn.1 TC0400004158.rn.1 RTA TC
          Rat Expression Set 230 1374450_at - -
          Rat Expression Set 230 1374450_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10863496 - -
          Rat Genome U34 Set rc_AA944855_at - -
          Rat Genome U34 Set rc_AI145501_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366878 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
          git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0