Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01422589_m1
          See other RNF152 GE Assays ›
          Gene Symbol:
          Rnf152
          Gene Name:
          ring finger protein 152
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 25186106 - 25262469 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01422589_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf152
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106305.1 NP_001099775.1 2 - 3 439 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001185.rn.1

          Additional Information:

          This assay is in the 5'UTR region of the NM_001106305.1 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          protein K48-linked ubiquitination
          ubiquitin-protein transferase activity
          zinc ion binding
          ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P776918 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P084578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656631 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148529 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122401 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77207 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52227 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90192 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100332 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102572 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110419 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76928 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102977 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P776918 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P084578 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P776918 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P084578 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P084578 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P776918 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P531962 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67904 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65448 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10766925 - -
          Affymetrix Rat Gene 1.0 ST Array 10766924 - -
          Affymetrix Rat Gene 1.1 ST Array 10766923 - -
          Rat Genome 230 2.0 Array 1379286_PM_at - -
          Rat Genome 230 2.0 Array 1379286_at - -
          Affymetrix Rat Clariom S Assay PSR1300007955.rn.1 TC1300001185.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300007954.rn.1 TC1300001185.rn.1 RTA TC
          Rat Expression Set 230 1379286_PM_at - -
          Rat Expression Set 230 1379286_at - -
          Affymetrix Rat Gene 1.0 ST Array 10766923 - -
          Rat Genome U34 Set rc_AI029086_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21846896 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0